Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17182543-17182814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR1
Entrez Gene ID
51816
Gene Name
cat eye syndrome chromosome region, candidate 1
Gene Aliases
ADA2, ADGF, IDGFL, PAN, SNEDS
UniGene
Hs.170310
Gene Chromosome Location
Chr.22: 17178790-17221854 on Build GRCh38
Transcripts
NM_001282225.1 NM_001282226.1 NM_001282227.1 NM_001282228.1 NM_001282229.1 NM_177405.2 XM_006724080.3 XM_011546133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4766845 c.1085G>T p.W362L Chr.22: 17182758-17182758 on Build GRCh38
6004788 c.1176C>T p.P392P Chr.22: 17182667-17182667 on Build GRCh38
5393115 c.1145_1146TC>GT p.I382S Chr.22: 17182697-17182698 on Build GRCh38
5393116 c.422_423TC>GT p.I141S Chr.22: 17182697-17182698 on Build GRCh38
4425497 c.441G>C p.L147F Chr.22: 17182679-17182679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749814301 Chr.22: 17182751-17182751 on Build GRCh38
rs763728425 Chr.22: 17182709-17182709 on Build GRCh38
rs577994684 Chr.22: 17182777-17182803 on Build GRCh38
rs759502049 Chr.22: 17182700-17182700 on Build GRCh38
rs543371262 Chr.22: 17182564-17182564 on Build GRCh38
rs746663293 Chr.22: 17182575-17182575 on Build GRCh38
rs901797579 Chr.22: 17182787-17182787 on Build GRCh38
rs764744427 Chr.22: 17182580-17182580 on Build GRCh38
rs757658203 Chr.22: 17182629-17182629 on Build GRCh38
rs774504121 Chr.22: 17182679-17182679 on Build GRCh38
rs756616828 Chr.22: 17182739-17182739 on Build GRCh38
rs750824133 Chr.22: 17182738-17182738 on Build GRCh38
rs752313204 Chr.22: 17182595-17182595 on Build GRCh38
rs769654299 Chr.22: 17182784-17182784 on Build GRCh38
rs981010876 Chr.22: 17182590-17182590 on Build GRCh38
rs772238679 Chr.22: 17182771-17182771 on Build GRCh38
rs776302859 Chr.22: 17182697-17182697 on Build GRCh38
rs749413678 Chr.22: 17182671-17182671 on Build GRCh38
rs367563832 Chr.22: 17182667-17182667 on Build GRCh38
rs749657410 Chr.22: 17182603-17182603 on Build GRCh38
rs772137894 Chr.22: 17182594-17182594 on Build GRCh38
rs755007390 Chr.22: 17182758-17182758 on Build GRCh38
rs745689865 Chr.22: 17182772-17182772 on Build GRCh38
rs373692780 Chr.22: 17182600-17182600 on Build GRCh38
rs765154508 Chr.22: 17182704-17182704 on Build GRCh38
rs910212904 Chr.22: 17182731-17182731 on Build GRCh38
rs777618863 Chr.22: 17182618-17182618 on Build GRCh38
rs770689762 Chr.22: 17182696-17182696 on Build GRCh38
rs372341852 Chr.22: 17182672-17182672 on Build GRCh38
rs780182069 Chr.22: 17182670-17182670 on Build GRCh38
rs770560241 Chr.22: 17182598-17182599 on Build GRCh38
rs751192866 Chr.22: 17182723-17182723 on Build GRCh38
rs767913207 Chr.22: 17182737-17182737 on Build GRCh38
rs972669448 Chr.22: 17182665-17182665 on Build GRCh38
rs781416927 Chr.22: 17182648-17182648 on Build GRCh38
rs748309664 Chr.22: 17182765-17182765 on Build GRCh38
rs960845017 Chr.22: 17182640-17182640 on Build GRCh38
rs17854705 Chr.22: 17182663-17182663 on Build GRCh38
rs759395875 Chr.22: 17182578-17182578 on Build GRCh38
rs780583418 Chr.22: 17182744-17182744 on Build GRCh38
rs375443506 Chr.22: 17182725-17182725 on Build GRCh38
rs561591791 Chr.22: 17182708-17182708 on Build GRCh38
rs928239647 Chr.22: 17182592-17182592 on Build GRCh38
rs762051665 Chr.22: 17182688-17182688 on Build GRCh38
rs370448970 Chr.22: 17182576-17182576 on Build GRCh38
rs780690814 Chr.22: 17182703-17182704 on Build GRCh38
rs751280698 Chr.22: 17182620-17182620 on Build GRCh38
rs746332176 Chr.22: 17182666-17182666 on Build GRCh38
rs778965407 Chr.22: 17182763-17182763 on Build GRCh38
rs766108390 Chr.22: 17182569-17182569 on Build GRCh38

View Full Product Details