Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44106650-44106907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100379224
Entrez Gene ID
100379224
Gene Name
uncharacterized LOC100379224
Gene Aliases
-
UniGene
Hs.585869
Gene Chromosome Location
Chr.19: 44105339-44113183 on Build GRCh38
Transcripts
NR_033341.1 NM_001321645.1 NM_013398.3 XM_017027261.1

Gene Symbol
ZNF224
Entrez Gene ID
7767
Gene Name
zinc finger protein 224
Gene Aliases
BMZF-2, BMZF2, KOX22, ZNF233, ZNF255, ZNF27
UniGene
Hs.585869
Gene Chromosome Location
Chr.19: 44094329-44109831 on Build GRCh38
Transcripts
NR_033341.1 NM_001321645.1 NM_013398.3 XM_017027261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1233708 c.490G>T p.D164Y Chr.19: 44106650-44106650 on Build GRCh38
474872 c.611A>G p.Y204C Chr.19: 44106771-44106771 on Build GRCh38
3289830 c.692C>A p.P231Q Chr.19: 44106852-44106852 on Build GRCh38
3289828 c.660A>G p.Q220Q Chr.19: 44106820-44106820 on Build GRCh38
6828663 c.603G>T p.E201D Chr.19: 44106763-44106763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370083464 Chr.19: 44106734-44106734 on Build GRCh38
rs748361208 Chr.19: 44106715-44106715 on Build GRCh38
rs777985241 Chr.19: 44106852-44106852 on Build GRCh38
rs756322847 Chr.19: 44106847-44106847 on Build GRCh38
rs779818283 Chr.19: 44106858-44106858 on Build GRCh38
rs768558299 Chr.19: 44106862-44106862 on Build GRCh38
rs775350960 Chr.19: 44106735-44106735 on Build GRCh38
rs1035599125 Chr.19: 44106861-44106861 on Build GRCh38
rs751692066 Chr.19: 44106706-44106706 on Build GRCh38
rs775013103 Chr.19: 44106683-44106683 on Build GRCh38
rs776173519 Chr.19: 44106771-44106771 on Build GRCh38
rs564513130 Chr.19: 44106880-44106880 on Build GRCh38
rs761342925 Chr.19: 44106869-44106869 on Build GRCh38
rs769175494 Chr.19: 44106724-44106726 on Build GRCh38
rs770039740 Chr.19: 44106717-44106717 on Build GRCh38
rs768220697 Chr.19: 44106755-44106755 on Build GRCh38
rs749350252 Chr.19: 44106713-44106714 on Build GRCh38
rs764894805 Chr.19: 44106781-44106781 on Build GRCh38
rs778598200 Chr.19: 44106726-44106726 on Build GRCh38
rs114131326 Chr.19: 44106817-44106817 on Build GRCh38
rs762243777 Chr.19: 44106757-44106759 on Build GRCh38
rs979267842 Chr.19: 44106794-44106794 on Build GRCh38
rs946547322 Chr.19: 44106765-44106765 on Build GRCh38
rs551748891 Chr.19: 44106704-44106704 on Build GRCh38
rs372573390 Chr.19: 44106687-44106687 on Build GRCh38
rs773611810 Chr.19: 44106828-44106829 on Build GRCh38
rs370833656 Chr.19: 44106829-44106829 on Build GRCh38
rs181803845 Chr.19: 44106820-44106820 on Build GRCh38
rs374131543 Chr.19: 44106785-44106785 on Build GRCh38
rs72480796 Chr.19: 44106690-44106690 on Build GRCh38
rs201293616 Chr.19: 44106822-44106822 on Build GRCh38
rs755047207 Chr.19: 44106710-44106710 on Build GRCh38
rs199772345 Chr.19: 44106870-44106870 on Build GRCh38
rs767716633 Chr.19: 44106812-44106813 on Build GRCh38
rs760594450 Chr.19: 44106749-44106749 on Build GRCh38
rs745637278 Chr.19: 44106731-44106731 on Build GRCh38
rs781419928 Chr.19: 44106712-44106712 on Build GRCh38
rs774681266 Chr.19: 44106739-44106741 on Build GRCh38
rs150633303 Chr.19: 44106689-44106689 on Build GRCh38
rs375784428 Chr.19: 44106691-44106691 on Build GRCh38
rs745665474 Chr.19: 44106853-44106853 on Build GRCh38
rs149287683 Chr.19: 44106700-44106700 on Build GRCh38
rs541964542 Chr.19: 44106782-44106782 on Build GRCh38
rs145285177 Chr.19: 44106860-44106860 on Build GRCh38
rs144310628 Chr.19: 44106775-44106775 on Build GRCh38
rs750107605 Chr.19: 44106744-44106745 on Build GRCh38
rs776026763 Chr.19: 44106865-44106865 on Build GRCh38
rs73562196 Chr.19: 44106801-44106801 on Build GRCh38

View Full Product Details