Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189694892-189695153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
UniGene
Hs.60772
Gene Chromosome Location
Chr.2: 189674292-189763059 on Build GRCh38
Transcripts
NM_144708.3 XM_011510673.2 XM_011510674.1 XM_011510675.2 XM_011510676.2 XM_011510677.2 XM_011510679.2 XM_011510680.1 XM_011510681.1 XM_011510682.2 XM_011510685.2 XM_011510686.2 XM_011510687.2 XM_011510688.1 XM_011510689.2 XM_011510691.2 XM_011510692.2 XM_011510693.2 XM_011510694.2 XM_017003413.1 XM_017003414.1 XM_017003415.1 XM_017003416.1 XM_017003417.1 XM_017003418.1 XM_017003419.1 XM_017003420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3838143 c.1426G>A p.D476N Chr.2: 189695099-189695099 on Build GRCh38
2712265 c.1423A>T p.T475S Chr.2: 189695096-189695096 on Build GRCh38
4661627 c.1168A>C p.N390H Chr.2: 189695054-189695054 on Build GRCh38
1014037 c.1105A>C p.I369L Chr.2: 189694991-189694991 on Build GRCh38
5452514 c.1107C>G p.I369M Chr.2: 189694993-189694993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138942918 Chr.2: 189695005-189695005 on Build GRCh38
rs772413468 Chr.2: 189694970-189694970 on Build GRCh38
rs755051839 Chr.2: 189695107-189695107 on Build GRCh38
rs530882331 Chr.2: 189695040-189695040 on Build GRCh38
rs574387659 Chr.2: 189695015-189695015 on Build GRCh38
rs781189421 Chr.2: 189694937-189694937 on Build GRCh38
rs141464406 Chr.2: 189695120-189695120 on Build GRCh38
rs753795809 Chr.2: 189695038-189695038 on Build GRCh38
rs758537604 Chr.2: 189694958-189694958 on Build GRCh38
rs762527864 Chr.2: 189694989-189694989 on Build GRCh38
rs949631002 Chr.2: 189695081-189695081 on Build GRCh38
rs763763301 Chr.2: 189694994-189694994 on Build GRCh38
rs768257928 Chr.2: 189695113-189695113 on Build GRCh38
rs767384825 Chr.2: 189695016-189695016 on Build GRCh38
rs780237969 Chr.2: 189695026-189695026 on Build GRCh38
rs761057452 Chr.2: 189694977-189694977 on Build GRCh38
rs958767220 Chr.2: 189695051-189695051 on Build GRCh38
rs750360745 Chr.2: 189694940-189694940 on Build GRCh38
rs772321638 Chr.2: 189695042-189695042 on Build GRCh38
rs746419356 Chr.2: 189695095-189695095 on Build GRCh38
rs750261011 Chr.2: 189695023-189695023 on Build GRCh38
rs966845986 Chr.2: 189694981-189694981 on Build GRCh38
rs780323928 Chr.2: 189694957-189694958 on Build GRCh38
rs917249832 Chr.2: 189695086-189695086 on Build GRCh38
rs773618111 Chr.2: 189694971-189694971 on Build GRCh38
rs777188349 Chr.2: 189695094-189695094 on Build GRCh38
rs991512747 Chr.2: 189695065-189695065 on Build GRCh38
rs779114563 Chr.2: 189694965-189694965 on Build GRCh38
rs771273946 Chr.2: 189695088-189695088 on Build GRCh38
rs779954461 Chr.2: 189694956-189694956 on Build GRCh38
rs534488091 Chr.2: 189694960-189694960 on Build GRCh38
rs747194558 Chr.2: 189695038-189695040 on Build GRCh38
rs115747871 Chr.2: 189694986-189694986 on Build GRCh38
rs754124669 Chr.2: 189695034-189695034 on Build GRCh38
rs755952696 Chr.2: 189695027-189695027 on Build GRCh38
rs747424329 Chr.2: 189695061-189695061 on Build GRCh38
rs780056581 Chr.2: 189695028-189695028 on Build GRCh38
rs768706610 Chr.2: 189694961-189694961 on Build GRCh38
rs778073997 Chr.2: 189695043-189695043 on Build GRCh38
rs998385447 Chr.2: 189694941-189694941 on Build GRCh38
rs971340594 Chr.2: 189695092-189695092 on Build GRCh38
rs769023694 Chr.2: 189695040-189695040 on Build GRCh38
rs748438429 Chr.2: 189694966-189694966 on Build GRCh38
rs916797288 Chr.2: 189695054-189695054 on Build GRCh38
rs756112105 Chr.2: 189694955-189694955 on Build GRCh38
rs756481102 Chr.2: 189694993-189694993 on Build GRCh38
rs1033525509 Chr.2: 189695022-189695022 on Build GRCh38

View Full Product Details