Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4207377-4207602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
UniGene
Hs.197872
Gene Chromosome Location
Chr.19: 4182618-4224814 on Build GRCh38
Transcripts
NM_133475.1 XM_011527756.2 XM_011527757.2 XM_011527758.2 XM_011527761.1 XM_011527764.2 XM_011527765.2 XM_017026438.1 XM_017026439.1 XM_017026440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7119175 c.898C>G p.Q300E Chr.19: 4207591-4207591 on Build GRCh38
4766203 c.295G>A p.A99T Chr.19: 4207582-4207582 on Build GRCh38
3389049 c.269G>A p.R90H Chr.19: 4207556-4207556 on Build GRCh38
3227176 c.214-1G>A p.? Chr.19: 4207500-4207500 on Build GRCh38
3227174 c.538-1G>A p.? Chr.19: 4207500-4207500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761109990 Chr.19: 4207504-4207504 on Build GRCh38
rs926094618 Chr.19: 4207451-4207451 on Build GRCh38
rs995119810 Chr.19: 4207563-4207563 on Build GRCh38
rs980377303 Chr.19: 4207401-4207402 on Build GRCh38
rs373784186 Chr.19: 4207467-4207467 on Build GRCh38
rs897243364 Chr.19: 4207570-4207570 on Build GRCh38
rs769652374 Chr.19: 4207564-4207564 on Build GRCh38
rs201865660 Chr.19: 4207579-4207579 on Build GRCh38
rs770905572 Chr.19: 4207577-4207577 on Build GRCh38
rs777530052 Chr.19: 4207566-4207566 on Build GRCh38
rs372390380 Chr.19: 4207507-4207507 on Build GRCh38
rs940817350 Chr.19: 4207551-4207551 on Build GRCh38
rs1040033990 Chr.19: 4207477-4207477 on Build GRCh38
rs935922484 Chr.19: 4207413-4207413 on Build GRCh38
rs1004754570 Chr.19: 4207541-4207541 on Build GRCh38
rs767260203 Chr.19: 4207491-4207491 on Build GRCh38
rs749191233 Chr.19: 4207576-4207576 on Build GRCh38
rs757375399 Chr.19: 4207519-4207519 on Build GRCh38
rs774126496 Chr.19: 4207426-4207426 on Build GRCh38
rs762399811 Chr.19: 4207454-4207454 on Build GRCh38
rs781712639 Chr.19: 4207489-4207489 on Build GRCh38
rs146188452 Chr.19: 4207556-4207556 on Build GRCh38
rs367658584 Chr.19: 4207478-4207478 on Build GRCh38
rs758685758 Chr.19: 4207547-4207547 on Build GRCh38
rs750585495 Chr.19: 4207532-4207532 on Build GRCh38
rs376699397 Chr.19: 4207581-4207581 on Build GRCh38
rs551206615 Chr.19: 4207407-4207407 on Build GRCh38
rs775226297 Chr.19: 4207583-4207583 on Build GRCh38
rs915618710 Chr.19: 4207459-4207459 on Build GRCh38
rs773214618 Chr.19: 4207452-4207452 on Build GRCh38
rs764453707 Chr.19: 4207506-4207506 on Build GRCh38
rs557303969 Chr.19: 4207436-4207437 on Build GRCh38
rs765903350 Chr.19: 4207455-4207455 on Build GRCh38
rs771557936 Chr.19: 4207582-4207582 on Build GRCh38
rs753000485 Chr.19: 4207494-4207494 on Build GRCh38
rs757745785 Chr.19: 4207483-4207485 on Build GRCh38
rs779111839 Chr.19: 4207523-4207523 on Build GRCh38
rs991108013 Chr.19: 4207445-4207445 on Build GRCh38
rs1045514788 Chr.19: 4207497-4207497 on Build GRCh38
rs371527951 Chr.19: 4207555-4207555 on Build GRCh38

View Full Product Details