Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115962468-115962729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LVRN
Entrez Gene ID
206338
Gene Name
laeverin
Gene Aliases
APQ, AQPEP, TAQPEP
UniGene
Hs.556880
Gene Chromosome Location
Chr.5: 115962440-116027602 on Build GRCh38
Transcripts
NM_173800.4 XM_011543263.2 XM_011543264.2 XM_017009215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4337780 c.7delC p.P4fs*8 Chr.5: 115962624-115962624 on Build GRCh38
1130961 c.64C>T p.L22L Chr.5: 115962681-115962681 on Build GRCh38
7082909 c.40G>T p.A14S Chr.5: 115962657-115962657 on Build GRCh38
6602645 c.111C>T p.Y37Y Chr.5: 115962728-115962728 on Build GRCh38
4127256 c.99C>T p.L33L Chr.5: 115962716-115962716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540265831 Chr.5: 115962646-115962646 on Build GRCh38
rs747042469 Chr.5: 115962588-115962588 on Build GRCh38
rs780442231 Chr.5: 115962655-115962655 on Build GRCh38
rs9326980 Chr.5: 115962535-115962535 on Build GRCh38
rs773316192 Chr.5: 115962701-115962701 on Build GRCh38
rs539685824 Chr.5: 115962644-115962644 on Build GRCh38
rs964167464 Chr.5: 115962523-115962523 on Build GRCh38
rs746331087 Chr.5: 115962653-115962653 on Build GRCh38
rs866059041 Chr.5: 115962649-115962649 on Build GRCh38
rs779289740 Chr.5: 115962568-115962568 on Build GRCh38
rs940733937 Chr.5: 115962519-115962519 on Build GRCh38
rs922687018 Chr.5: 115962543-115962543 on Build GRCh38
rs752878928 Chr.5: 115962616-115962616 on Build GRCh38
rs766397285 Chr.5: 115962708-115962708 on Build GRCh38
rs761854251 Chr.5: 115962554-115962554 on Build GRCh38
rs1029667108 Chr.5: 115962691-115962691 on Build GRCh38
rs878974207 Chr.5: 115962594-115962594 on Build GRCh38
rs759884993 Chr.5: 115962530-115962530 on Build GRCh38
rs757221581 Chr.5: 115962645-115962645 on Build GRCh38
rs1014344903 Chr.5: 115962591-115962591 on Build GRCh38
rs748241080 Chr.5: 115962486-115962486 on Build GRCh38
rs552871898 Chr.5: 115962667-115962667 on Build GRCh38
rs200890347 Chr.5: 115962641-115962641 on Build GRCh38
rs771657133 Chr.5: 115962689-115962689 on Build GRCh38
rs186797849 Chr.5: 115962506-115962506 on Build GRCh38
rs36066288 Chr.5: 115962666-115962667 on Build GRCh38
rs560631931 Chr.5: 115962518-115962518 on Build GRCh38
rs201363777 Chr.5: 115962614-115962614 on Build GRCh38
rs773615364 Chr.5: 115962504-115962504 on Build GRCh38
rs367912672 Chr.5: 115962654-115962654 on Build GRCh38
rs781664236 Chr.5: 115962596-115962596 on Build GRCh38
rs750325985 Chr.5: 115962626-115962626 on Build GRCh38
rs934042179 Chr.5: 115962556-115962556 on Build GRCh38
rs983155211 Chr.5: 115962558-115962558 on Build GRCh38
rs765325913 Chr.5: 115962528-115962529 on Build GRCh38
rs894173513 Chr.5: 115962582-115962582 on Build GRCh38
rs767342846 Chr.5: 115962555-115962555 on Build GRCh38
rs775152167 Chr.5: 115962608-115962608 on Build GRCh38
rs750309575 Chr.5: 115962559-115962559 on Build GRCh38
rs374591604 Chr.5: 115962605-115962605 on Build GRCh38
rs953618097 Chr.5: 115962694-115962694 on Build GRCh38
rs746238759 Chr.5: 115962598-115962598 on Build GRCh38
rs760570873 Chr.5: 115962628-115962628 on Build GRCh38
rs760736455 Chr.5: 115962702-115962702 on Build GRCh38
rs139985376 Chr.5: 115962578-115962578 on Build GRCh38
rs564621239 Chr.5: 115962485-115962485 on Build GRCh38
rs761223132 Chr.5: 115962625-115962625 on Build GRCh38
rs931968616 Chr.5: 115962507-115962507 on Build GRCh38
rs771529867 Chr.5: 115962595-115962595 on Build GRCh38
rs377593017 Chr.5: 115962547-115962547 on Build GRCh38
rs371899368 Chr.5: 115962635-115962635 on Build GRCh38
rs769506577 Chr.5: 115962533-115962533 on Build GRCh38
rs777106458 Chr.5: 115962528-115962528 on Build GRCh38
rs755904111 Chr.5: 115962564-115962564 on Build GRCh38
rs547197649 Chr.5: 115962509-115962509 on Build GRCh38
rs773969321 Chr.5: 115962624-115962624 on Build GRCh38
rs10062297 Chr.5: 115962681-115962681 on Build GRCh38
rs773807537 Chr.5: 115962669-115962669 on Build GRCh38
rs777948678 Chr.5: 115962580-115962580 on Build GRCh38
rs768166990 Chr.5: 115962619-115962619 on Build GRCh38
rs749598779 Chr.5: 115962656-115962656 on Build GRCh38
rs538402262 Chr.5: 115962541-115962541 on Build GRCh38
rs763616917 Chr.5: 115962542-115962542 on Build GRCh38
rs548268324 Chr.5: 115962576-115962576 on Build GRCh38
rs772134876 Chr.5: 115962487-115962487 on Build GRCh38

View Full Product Details