Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85865037-85865310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC83
Entrez Gene ID
220047
Gene Name
coiled-coil domain containing 83
Gene Aliases
CT148, HSD9
UniGene
Hs.567774
Gene Chromosome Location
Chr.11: 85855066-85920021 on Build GRCh38
Transcripts
NM_001286159.1 NM_173556.4 XM_011544839.2 XM_011544840.2 XM_011544841.1 XM_011544842.1 XM_011544843.1 XM_017017354.1 XM_017017355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5373746 c.94C>A p.Q32K Chr.11: 85865217-85865217 on Build GRCh38
3454088 c.33T>C p.D11D Chr.11: 85865156-85865156 on Build GRCh38
5477860 c.30G>T p.K10N Chr.11: 85865153-85865153 on Build GRCh38
2221502 c.67C>T p.P23S Chr.11: 85865190-85865190 on Build GRCh38
2221500 c.12A>G p.S4S Chr.11: 85865135-85865135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762309041 Chr.11: 85865074-85865074 on Build GRCh38
rs745421600 Chr.11: 85865162-85865162 on Build GRCh38
rs765143778 Chr.11: 85865248-85865248 on Build GRCh38
rs778858952 Chr.11: 85865099-85865099 on Build GRCh38
rs201205802 Chr.11: 85865077-85865077 on Build GRCh38
rs374138415 Chr.11: 85865239-85865239 on Build GRCh38
rs752647624 Chr.11: 85865252-85865252 on Build GRCh38
rs199576043 Chr.11: 85865154-85865154 on Build GRCh38
rs777464406 Chr.11: 85865267-85865267 on Build GRCh38
rs779482361 Chr.11: 85865075-85865075 on Build GRCh38
rs773442206 Chr.11: 85865177-85865177 on Build GRCh38
rs753726842 Chr.11: 85865096-85865096 on Build GRCh38
rs142636291 Chr.11: 85865166-85865166 on Build GRCh38
rs200674793 Chr.11: 85865163-85865163 on Build GRCh38
rs747929865 Chr.11: 85865102-85865102 on Build GRCh38
rs397942737 Chr.11: 85865073-85865074 on Build GRCh38
rs767776100 Chr.11: 85865167-85865167 on Build GRCh38
rs981052341 Chr.11: 85865197-85865197 on Build GRCh38
rs746648432 Chr.11: 85865137-85865137 on Build GRCh38
rs755113636 Chr.11: 85865245-85865245 on Build GRCh38
rs900009029 Chr.11: 85865069-85865069 on Build GRCh38
rs770438448 Chr.11: 85865138-85865138 on Build GRCh38
rs760829831 Chr.11: 85865187-85865187 on Build GRCh38
rs758147943 Chr.11: 85865257-85865257 on Build GRCh38
rs777428015 Chr.11: 85865116-85865116 on Build GRCh38
rs199934074 Chr.11: 85865165-85865165 on Build GRCh38
rs11389838 Chr.11: 85865070-85865071 on Build GRCh38
rs758182215 Chr.11: 85865104-85865104 on Build GRCh38
rs753880351 Chr.11: 85865241-85865241 on Build GRCh38
rs957573926 Chr.11: 85865079-85865079 on Build GRCh38
rs36093844 Chr.11: 85865084-85865084 on Build GRCh38
rs754883549 Chr.11: 85865097-85865097 on Build GRCh38

View Full Product Details