Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151148549-151148820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBX1
Entrez Gene ID
2636
Gene Name
gastrulation brain homeobox 1
Gene Aliases
Huh-17
UniGene
Gene Chromosome Location
Chr.7: 151148009-151167548 on Build GRCh38
Transcripts
NM_001098834.2 XM_017011964.1 XM_017011965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4797556 c.1024C>T p.P342S Chr.7: 151148657-151148657 on Build GRCh38
6486642 c.988G>A p.E330K Chr.7: 151148693-151148693 on Build GRCh38
6177182 c.1083C>T p.A361A Chr.7: 151148598-151148598 on Build GRCh38
4686572 c.1055G>T p.S352I Chr.7: 151148626-151148626 on Build GRCh38
3636331 c.940G>A p.A314T Chr.7: 151148741-151148741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762691879 Chr.7: 151148793-151148793 on Build GRCh38
rs201566761 Chr.7: 151148635-151148635 on Build GRCh38
rs199515474 Chr.7: 151148701-151148701 on Build GRCh38
rs760715412 Chr.7: 151148688-151148688 on Build GRCh38
rs376045645 Chr.7: 151148709-151148709 on Build GRCh38
rs758029438 Chr.7: 151148728-151148728 on Build GRCh38
rs751823436 Chr.7: 151148576-151148576 on Build GRCh38
rs747866558 Chr.7: 151148600-151148600 on Build GRCh38
rs1020827991 Chr.7: 151148790-151148790 on Build GRCh38
rs766876227 Chr.7: 151148691-151148691 on Build GRCh38
rs757328162 Chr.7: 151148579-151148579 on Build GRCh38
rs750469482 Chr.7: 151148590-151148590 on Build GRCh38
rs770855143 Chr.7: 151148615-151148615 on Build GRCh38
rs774547279 Chr.7: 151148649-151148649 on Build GRCh38
rs767697983 Chr.7: 151148584-151148584 on Build GRCh38
rs202046564 Chr.7: 151148630-151148630 on Build GRCh38
rs373562908 Chr.7: 151148745-151148745 on Build GRCh38
rs368877994 Chr.7: 151148729-151148729 on Build GRCh38
rs762023838 Chr.7: 151148660-151148660 on Build GRCh38
rs776601538 Chr.7: 151148629-151148629 on Build GRCh38
rs767661301 Chr.7: 151148672-151148672 on Build GRCh38
rs746963346 Chr.7: 151148597-151148597 on Build GRCh38
rs779363227 Chr.7: 151148707-151148707 on Build GRCh38
rs753116482 Chr.7: 151148718-151148718 on Build GRCh38
rs770390765 Chr.7: 151148741-151148741 on Build GRCh38
rs563201738 Chr.7: 151148705-151148705 on Build GRCh38
rs75176255 Chr.7: 151148766-151148766 on Build GRCh38
rs964388952 Chr.7: 151148768-151148768 on Build GRCh38
rs907006933 Chr.7: 151148659-151148659 on Build GRCh38
rs1047351798 Chr.7: 151148632-151148632 on Build GRCh38
rs376566770 Chr.7: 151148596-151148596 on Build GRCh38
rs777271750 Chr.7: 151148607-151148607 on Build GRCh38
rs756600488 Chr.7: 151148595-151148595 on Build GRCh38
rs745869295 Chr.7: 151148746-151148746 on Build GRCh38
rs1045636166 Chr.7: 151148639-151148639 on Build GRCh38
rs778703161 Chr.7: 151148599-151148599 on Build GRCh38
rs899566288 Chr.7: 151148694-151148694 on Build GRCh38
rs746533652 Chr.7: 151148613-151148613 on Build GRCh38
rs770948033 Chr.7: 151148642-151148642 on Build GRCh38
rs941507279 Chr.7: 151148637-151148637 on Build GRCh38
rs978544871 Chr.7: 151148580-151148580 on Build GRCh38
rs200609477 Chr.7: 151148792-151148792 on Build GRCh38
rs772310395 Chr.7: 151148798-151148798 on Build GRCh38
rs776675913 Chr.7: 151148794-151148794 on Build GRCh38
rs750465302 Chr.7: 151148687-151148687 on Build GRCh38
rs775219988 Chr.7: 151148769-151148769 on Build GRCh38
rs201417319 Chr.7: 151148799-151148799 on Build GRCh38

View Full Product Details