Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171635879-171636134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOC
Entrez Gene ID
4653
Gene Name
myocilin
Gene Aliases
GLC1A, GPOA, JOAG, JOAG1, TIGR
UniGene
Hs.436037
Gene Chromosome Location
Chr.1: 171635417-171652633 on Build GRCh38
Transcripts
NM_000261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5822724 c.1478A>T p.N493I Chr.1: 171635962-171635962 on Build GRCh38
5378429 c.1392C>T p.T464T Chr.1: 171636048-171636048 on Build GRCh38
5917287 c.1421G>A p.S474N Chr.1: 171636019-171636019 on Build GRCh38
354086 c.1491T>C p.Y497Y Chr.1: 171635949-171635949 on Build GRCh38
1320751 c.1407C>G p.N469K Chr.1: 171636033-171636033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755575564 Chr.1: 171636075-171636075 on Build GRCh38
rs1034625739 Chr.1: 171635919-171635919 on Build GRCh38
rs754248248 Chr.1: 171636057-171636057 on Build GRCh38
rs776513776 Chr.1: 171635977-171635977 on Build GRCh38
rs777445056 Chr.1: 171635968-171635968 on Build GRCh38
rs958675451 Chr.1: 171635923-171635923 on Build GRCh38
rs727504025 Chr.1: 171635949-171635949 on Build GRCh38
rs750800370 Chr.1: 171635921-171635921 on Build GRCh38
rs145977437 Chr.1: 171635941-171635941 on Build GRCh38
rs745712367 Chr.1: 171635982-171635982 on Build GRCh38
rs769863509 Chr.1: 171636052-171636053 on Build GRCh38
rs764112994 Chr.1: 171636014-171636014 on Build GRCh38
rs140967767 Chr.1: 171636106-171636106 on Build GRCh38
rs756472086 Chr.1: 171636034-171636034 on Build GRCh38
rs766884379 Chr.1: 171636036-171636036 on Build GRCh38
rs750791099 Chr.1: 171636031-171636031 on Build GRCh38
rs756630885 Chr.1: 171635933-171635933 on Build GRCh38
rs757893552 Chr.1: 171635946-171635946 on Build GRCh38
rs576458696 Chr.1: 171636083-171636083 on Build GRCh38
rs747543321 Chr.1: 171636022-171636025 on Build GRCh38
rs771122834 Chr.1: 171636032-171636032 on Build GRCh38
rs2234929 Chr.1: 171635976-171635976 on Build GRCh38
rs145447699 Chr.1: 171636069-171636069 on Build GRCh38
rs1014299875 Chr.1: 171635961-171635961 on Build GRCh38
rs142680834 Chr.1: 171636104-171636104 on Build GRCh38
rs554235897 Chr.1: 171636028-171636028 on Build GRCh38
rs775380030 Chr.1: 171636112-171636112 on Build GRCh38
rs764272028 Chr.1: 171635900-171635900 on Build GRCh38
rs757499936 Chr.1: 171635905-171635905 on Build GRCh38
rs1024809888 Chr.1: 171635971-171635971 on Build GRCh38
rs760260934 Chr.1: 171636013-171636013 on Build GRCh38
rs761963923 Chr.1: 171636027-171636027 on Build GRCh38
rs746534441 Chr.1: 171636096-171636096 on Build GRCh38
rs74315331 Chr.1: 171636010-171636010 on Build GRCh38
rs767801045 Chr.1: 171635907-171635907 on Build GRCh38
rs769767738 Chr.1: 171635985-171635985 on Build GRCh38
rs767733168 Chr.1: 171635904-171635904 on Build GRCh38
rs74315332 Chr.1: 171636000-171636000 on Build GRCh38
rs776269268 Chr.1: 171635978-171635979 on Build GRCh38
rs572512491 Chr.1: 171636095-171636095 on Build GRCh38
rs763068244 Chr.1: 171635999-171635999 on Build GRCh38
rs774417217 Chr.1: 171636021-171636021 on Build GRCh38
rs756961599 Chr.1: 171636100-171636100 on Build GRCh38
rs868311329 Chr.1: 171636067-171636067 on Build GRCh38
rs376735175 Chr.1: 171635990-171635990 on Build GRCh38

View Full Product Details