Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121854608-121854853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPD
Entrez Gene ID
3242
Gene Name
4-hydroxyphenylpyruvate dioxygenase
Gene Aliases
4-HPPD, 4HPPD, GLOD3, HPPDASE, PPD
UniGene
Hs.2899
Gene Chromosome Location
Chr.12: 121839527-121888611 on Build GRCh38
Transcripts
NM_001171993.1 NM_002150.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209933 c.392T>C p.V131A Chr.12: 121854725-121854725 on Build GRCh38
3368692 c.334G>A p.E112K Chr.12: 121854783-121854783 on Build GRCh38
3870955 c.340G>A p.G114S Chr.12: 121854777-121854777 on Build GRCh38
1254380 c.337C>T p.R113W Chr.12: 121854780-121854780 on Build GRCh38
5618483 c.338G>A p.R113Q Chr.12: 121854779-121854779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776395434 Chr.12: 121854675-121854675 on Build GRCh38
rs757958827 Chr.12: 121854796-121854796 on Build GRCh38
rs375816927 Chr.12: 121854717-121854717 on Build GRCh38
rs545913399 Chr.12: 121854719-121854719 on Build GRCh38
rs761022712 Chr.12: 121854781-121854781 on Build GRCh38
rs779947081 Chr.12: 121854815-121854815 on Build GRCh38
rs200528285 Chr.12: 121854811-121854811 on Build GRCh38
rs751129052 Chr.12: 121854807-121854807 on Build GRCh38
rs775478030 Chr.12: 121854771-121854771 on Build GRCh38
rs746774715 Chr.12: 121854665-121854665 on Build GRCh38
rs763100741 Chr.12: 121854691-121854691 on Build GRCh38
rs113472814 Chr.12: 121854702-121854702 on Build GRCh38
rs969683124 Chr.12: 121854644-121854644 on Build GRCh38
rs777553334 Chr.12: 121854736-121854736 on Build GRCh38
rs746724701 Chr.12: 121854749-121854749 on Build GRCh38
rs368438884 Chr.12: 121854780-121854780 on Build GRCh38
rs1053332385 Chr.12: 121854801-121854801 on Build GRCh38
rs754907145 Chr.12: 121854728-121854728 on Build GRCh38
rs756970460 Chr.12: 121854757-121854757 on Build GRCh38
rs745676456 Chr.12: 121854812-121854812 on Build GRCh38
rs745739337 Chr.12: 121854762-121854762 on Build GRCh38
rs768775184 Chr.12: 121854774-121854774 on Build GRCh38
rs113807474 Chr.12: 121854701-121854701 on Build GRCh38
rs755963838 Chr.12: 121854814-121854814 on Build GRCh38
rs117839625 Chr.12: 121854786-121854786 on Build GRCh38
rs774512978 Chr.12: 121854823-121854823 on Build GRCh38
rs11833399 Chr.12: 121854779-121854779 on Build GRCh38
rs146428624 Chr.12: 121854685-121854685 on Build GRCh38
rs762065329 Chr.12: 121854704-121854704 on Build GRCh38
rs920320378 Chr.12: 121854746-121854746 on Build GRCh38
rs140788110 Chr.12: 121854703-121854703 on Build GRCh38
rs759554947 Chr.12: 121854679-121854679 on Build GRCh38
rs3741586 Chr.12: 121854640-121854640 on Build GRCh38
rs2620338 Chr.12: 121854650-121854650 on Build GRCh38
rs776410155 Chr.12: 121854666-121854666 on Build GRCh38
rs149180825 Chr.12: 121854761-121854761 on Build GRCh38
rs754452969 Chr.12: 121854795-121854795 on Build GRCh38
rs768685269 Chr.12: 121854818-121854818 on Build GRCh38
rs59741465 Chr.12: 121854772-121854772 on Build GRCh38
rs769734231 Chr.12: 121854765-121854765 on Build GRCh38
rs764298771 Chr.12: 121854692-121854692 on Build GRCh38
rs777643375 Chr.12: 121854662-121854662 on Build GRCh38
rs376148151 Chr.12: 121854817-121854817 on Build GRCh38
rs1048776123 Chr.12: 121854726-121854726 on Build GRCh38
rs61742674 Chr.12: 121854775-121854775 on Build GRCh38
rs1010875174 Chr.12: 121854648-121854648 on Build GRCh38
rs763736499 Chr.12: 121854804-121854804 on Build GRCh38
rs764783765 Chr.12: 121854720-121854720 on Build GRCh38
rs769624201 Chr.12: 121854639-121854639 on Build GRCh38
rs758046827 Chr.12: 121854734-121854734 on Build GRCh38
rs767968204 Chr.12: 121854714-121854714 on Build GRCh38

View Full Product Details