Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89686397-89686670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC8C
Entrez Gene ID
84230
Gene Name
leucine rich repeat containing 8 family member C
Gene Aliases
AD158, FAD158
UniGene
Hs.412836
Gene Chromosome Location
Chr.1: 89615810-89719535 on Build GRCh38
Transcripts
NM_032270.4 XM_006710960.3 XM_011542282.2 XM_011542283.2 XM_017002503.1 XM_017002504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
913046 c.25C>T p.Q9* Chr.1: 89686498-89686498 on Build GRCh38
2259172 c.57G>A p.V19V Chr.1: 89686530-89686530 on Build GRCh38
2259170 c.18A>T p.E6D Chr.1: 89686491-89686491 on Build GRCh38
6567129 c.26A>G p.Q9R Chr.1: 89686499-89686499 on Build GRCh38
3377304 c.31T>G p.S11A Chr.1: 89686504-89686504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112191990 Chr.1: 89686482-89686482 on Build GRCh38
rs749602831 Chr.1: 89686644-89686644 on Build GRCh38
rs371828858 Chr.1: 89686595-89686595 on Build GRCh38
rs200088710 Chr.1: 89686457-89686457 on Build GRCh38
rs781532253 Chr.1: 89686495-89686495 on Build GRCh38
rs898342612 Chr.1: 89686444-89686444 on Build GRCh38
rs766214229 Chr.1: 89686428-89686428 on Build GRCh38
rs777861840 Chr.1: 89686601-89686601 on Build GRCh38
rs757389941 Chr.1: 89686621-89686621 on Build GRCh38
rs752719047 Chr.1: 89686618-89686618 on Build GRCh38
rs770160931 Chr.1: 89686505-89686505 on Build GRCh38
rs780339248 Chr.1: 89686642-89686642 on Build GRCh38
rs751573376 Chr.1: 89686483-89686483 on Build GRCh38
rs754802346 Chr.1: 89686464-89686464 on Build GRCh38
rs541280497 Chr.1: 89686465-89686465 on Build GRCh38
rs768988693 Chr.1: 89686539-89686539 on Build GRCh38
rs774981080 Chr.1: 89686551-89686551 on Build GRCh38
rs780240679 Chr.1: 89686521-89686521 on Build GRCh38
rs746141407 Chr.1: 89686503-89686503 on Build GRCh38
rs114433988 Chr.1: 89686590-89686590 on Build GRCh38
rs768405271 Chr.1: 89686588-89686588 on Build GRCh38
rs771614537 Chr.1: 89686573-89686573 on Build GRCh38
rs757475640 Chr.1: 89686489-89686489 on Build GRCh38
rs140629995 Chr.1: 89686605-89686605 on Build GRCh38
rs753515695 Chr.1: 89686593-89686593 on Build GRCh38
rs74098169 Chr.1: 89686557-89686557 on Build GRCh38
rs760436810 Chr.1: 89686423-89686423 on Build GRCh38
rs367978975 Chr.1: 89686425-89686425 on Build GRCh38
rs371789599 Chr.1: 89686450-89686450 on Build GRCh38
rs912214743 Chr.1: 89686447-89686447 on Build GRCh38
rs756423537 Chr.1: 89686544-89686544 on Build GRCh38
rs750560887 Chr.1: 89686630-89686630 on Build GRCh38
rs1022882486 Chr.1: 89686480-89686480 on Build GRCh38
rs781370531 Chr.1: 89686626-89686626 on Build GRCh38
rs374852558 Chr.1: 89686525-89686525 on Build GRCh38
rs10082200 Chr.1: 89686587-89686587 on Build GRCh38
rs756340131 Chr.1: 89686641-89686641 on Build GRCh38

View Full Product Details