Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19262623-19262854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN4
Entrez Gene ID
404037
Gene Name
hyaluronan and proteoglycan link protein 4
Gene Aliases
BRAL2
UniGene
Hs.367829
Gene Chromosome Location
Chr.19: 19255641-19262787 on Build GRCh38
Transcripts
NM_023002.2

SNP Detail

SNP ID Chromosome Location
rs886745902 Chr.19: 19262787-19262787 on Build GRCh38
rs754562616 Chr.19: 19262781-19262781 on Build GRCh38
rs914107571 Chr.19: 19262780-19262780 on Build GRCh38
rs372876724 Chr.19: 19262785-19262785 on Build GRCh38
rs777021904 Chr.19: 19262729-19262729 on Build GRCh38
rs748697414 Chr.19: 19262708-19262708 on Build GRCh38
rs770004739 Chr.19: 19262740-19262740 on Build GRCh38
rs754508999 Chr.19: 19262707-19262707 on Build GRCh38
rs942521888 Chr.19: 19262741-19262741 on Build GRCh38
rs778326550 Chr.19: 19262722-19262722 on Build GRCh38
rs772525584 Chr.19: 19262709-19262709 on Build GRCh38
rs956720277 Chr.19: 19262672-19262672 on Build GRCh38
rs767404192 Chr.19: 19262750-19262750 on Build GRCh38
rs775364590 Chr.19: 19262746-19262746 on Build GRCh38
rs757084803 Chr.19: 19262689-19262689 on Build GRCh38
rs752677513 Chr.19: 19262745-19262746 on Build GRCh38
rs1005239703 Chr.19: 19262795-19262795 on Build GRCh38
rs767348383 Chr.19: 19262691-19262691 on Build GRCh38
rs753449731 Chr.19: 19262704-19262704 on Build GRCh38
rs531041176 Chr.19: 19262813-19262813 on Build GRCh38
rs775078166 Chr.19: 19262788-19262788 on Build GRCh38
rs750344573 Chr.19: 19262756-19262756 on Build GRCh38
rs760496163 Chr.19: 19262766-19262766 on Build GRCh38
rs965203918 Chr.19: 19262663-19262663 on Build GRCh38
rs140808775 Chr.19: 19262738-19262751 on Build GRCh38
rs550243655 Chr.19: 19262822-19262822 on Build GRCh38
rs759782265 Chr.19: 19262739-19262739 on Build GRCh38
rs766116011 Chr.19: 19262768-19262768 on Build GRCh38
rs758507798 Chr.19: 19262767-19262768 on Build GRCh38
rs991123567 Chr.19: 19262736-19262744 on Build GRCh38
rs1046982273 Chr.19: 19262786-19262786 on Build GRCh38
rs747377962 Chr.19: 19262724-19262724 on Build GRCh38
rs779724048 Chr.19: 19262702-19262702 on Build GRCh38
rs1038114292 Chr.19: 19262799-19262799 on Build GRCh38
rs377295631 Chr.19: 19262682-19262682 on Build GRCh38
rs181834483 Chr.19: 19262684-19262684 on Build GRCh38
rs753504837 Chr.19: 19262778-19262778 on Build GRCh38
rs953888222 Chr.19: 19262730-19262730 on Build GRCh38
rs1015458579 Chr.19: 19262654-19262654 on Build GRCh38
rs769364055 Chr.19: 19262705-19262705 on Build GRCh38
rs117397390 Chr.19: 19262673-19262673 on Build GRCh38
rs1002673569 Chr.19: 19262811-19262811 on Build GRCh38
rs759653922 Chr.19: 19262719-19262744 on Build GRCh38

View Full Product Details