Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29906721-29906964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPHD1
Entrez Gene ID
253982
Gene Name
aspartate beta-hydroxylase domain containing 1
Gene Aliases
-
UniGene
Hs.655761
Gene Chromosome Location
Chr.16: 29900392-29919860 on Build GRCh38
Transcripts
NM_178863.4 NR_110933.1 XM_011545783.2 XM_011545784.2 XM_017023104.1

Gene Symbol
KCTD13
Entrez Gene ID
253980
Gene Name
potassium channel tetramerization domain containing 13
Gene Aliases
BACURD1, FKSG86, PDIP1, POLDIP1, hBACURD1
UniGene
Hs.534590
Gene Chromosome Location
Chr.16: 29906335-29926247 on Build GRCh38
Transcripts
NM_178863.4 NR_110933.1 XM_011545783.2 XM_011545784.2 XM_017023104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692026 c.948C>T p.H316H Chr.16: 29906914-29906914 on Build GRCh38
4887850 c.916C>T p.R306C Chr.16: 29906946-29906946 on Build GRCh38
4978900 c.949G>A p.D317N Chr.16: 29906913-29906913 on Build GRCh38
969695 c.952G>A p.E318K Chr.16: 29906910-29906910 on Build GRCh38
6692021 c.902G>A p.R301Q Chr.16: 29906960-29906960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201785340 Chr.16: 29906850-29906850 on Build GRCh38
rs980884750 Chr.16: 29906806-29906806 on Build GRCh38
rs960888653 Chr.16: 29906840-29906840 on Build GRCh38
rs557468106 Chr.16: 29906763-29906763 on Build GRCh38
rs973373938 Chr.16: 29906907-29906907 on Build GRCh38
rs769549759 Chr.16: 29906877-29906877 on Build GRCh38
rs754709131 Chr.16: 29906772-29906772 on Build GRCh38
rs563953632 Chr.16: 29906914-29906914 on Build GRCh38
rs1022304792 Chr.16: 29906764-29906764 on Build GRCh38
rs372728677 Chr.16: 29906946-29906946 on Build GRCh38
rs371356818 Chr.16: 29906871-29906871 on Build GRCh38
rs541171011 Chr.16: 29906791-29906791 on Build GRCh38
rs574044040 Chr.16: 29906786-29906786 on Build GRCh38
rs184896100 Chr.16: 29906860-29906860 on Build GRCh38
rs954127010 Chr.16: 29906760-29906760 on Build GRCh38
rs773039788 Chr.16: 29906911-29906911 on Build GRCh38
rs753123512 Chr.16: 29906927-29906927 on Build GRCh38
rs561754578 Chr.16: 29906783-29906783 on Build GRCh38
rs371170171 Chr.16: 29906913-29906913 on Build GRCh38
rs1034910953 Chr.16: 29906943-29906953 on Build GRCh38
rs970964740 Chr.16: 29906795-29906795 on Build GRCh38
rs907551420 Chr.16: 29906930-29906930 on Build GRCh38
rs780033627 Chr.16: 29906837-29906837 on Build GRCh38
rs990982984 Chr.16: 29906854-29906854 on Build GRCh38
rs746975483 Chr.16: 29906856-29906856 on Build GRCh38
rs771799055 Chr.16: 29906822-29906822 on Build GRCh38
rs978379667 Chr.16: 29906773-29906773 on Build GRCh38
rs200627102 Chr.16: 29906864-29906864 on Build GRCh38

View Full Product Details