Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55876576-55876849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP1
Entrez Gene ID
2202
Gene Name
EGF containing fibulin like extracellular matrix protein 1
Gene Aliases
DHRD, DRAD, FBLN3, FBNL, FIBL-3, MLVT, MTLV, S1-5
UniGene
Hs.732348
Gene Chromosome Location
Chr.2: 55865962-55924163 on Build GRCh38
Transcripts
NM_001039348.2 NM_001039349.2 XM_005264205.4 XM_017003586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761081 c.769G>T p.E257* Chr.2: 55876734-55876734 on Build GRCh38
3743844 c.824T>C p.F275S Chr.2: 55876679-55876679 on Build GRCh38
5476390 c.851A>G p.E284G Chr.2: 55876652-55876652 on Build GRCh38
1021907 c.873C>T p.N291N Chr.2: 55876630-55876630 on Build GRCh38
1408720 c.807C>T p.Y269Y Chr.2: 55876696-55876696 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370306621 Chr.2: 55876615-55876615 on Build GRCh38
rs762971862 Chr.2: 55876758-55876758 on Build GRCh38
rs138120227 Chr.2: 55876696-55876696 on Build GRCh38
rs148488868 Chr.2: 55876718-55876718 on Build GRCh38
rs751482730 Chr.2: 55876754-55876755 on Build GRCh38
rs796715832 Chr.2: 55876757-55876758 on Build GRCh38
rs3838530 Chr.2: 55876756-55876757 on Build GRCh38
rs868355677 Chr.2: 55876658-55876658 on Build GRCh38
rs748893900 Chr.2: 55876769-55876772 on Build GRCh38
rs763898876 Chr.2: 55876762-55876762 on Build GRCh38
rs751492340 Chr.2: 55876765-55876765 on Build GRCh38
rs779945400 Chr.2: 55876753-55876753 on Build GRCh38
rs1005784311 Chr.2: 55876626-55876626 on Build GRCh38
rs780959367 Chr.2: 55876771-55876771 on Build GRCh38
rs1027805547 Chr.2: 55876795-55876796 on Build GRCh38
rs769930393 Chr.2: 55876614-55876614 on Build GRCh38
rs752587852 Chr.2: 55876820-55876820 on Build GRCh38
rs762128597 Chr.2: 55876714-55876714 on Build GRCh38
rs1034615733 Chr.2: 55876642-55876642 on Build GRCh38
rs757101511 Chr.2: 55876767-55876767 on Build GRCh38
rs776487850 Chr.2: 55876720-55876720 on Build GRCh38
rs200161978 Chr.2: 55876787-55876788 on Build GRCh38
rs201304043 Chr.2: 55876757-55876758 on Build GRCh38
rs942713794 Chr.2: 55876799-55876799 on Build GRCh38
rs749283148 Chr.2: 55876790-55876790 on Build GRCh38
rs759260191 Chr.2: 55876724-55876724 on Build GRCh38
rs569812649 Chr.2: 55876756-55876756 on Build GRCh38
rs764846730 Chr.2: 55876740-55876740 on Build GRCh38
rs769121277 Chr.2: 55876757-55876762 on Build GRCh38
rs372765157 Chr.2: 55876774-55876774 on Build GRCh38
rs925213696 Chr.2: 55876643-55876643 on Build GRCh38
rs756268360 Chr.2: 55876776-55876776 on Build GRCh38
rs538492843 Chr.2: 55876805-55876805 on Build GRCh38
rs752091023 Chr.2: 55876750-55876750 on Build GRCh38
rs749818060 Chr.2: 55876627-55876627 on Build GRCh38
rs768998905 Chr.2: 55876651-55876651 on Build GRCh38
rs758279735 Chr.2: 55876746-55876746 on Build GRCh38
rs777418265 Chr.2: 55876758-55876759 on Build GRCh38
rs778701916 Chr.2: 55876787-55876787 on Build GRCh38
rs754044580 Chr.2: 55876609-55876610 on Build GRCh38
rs911131323 Chr.2: 55876797-55876797 on Build GRCh38
rs1024726454 Chr.2: 55876746-55876746 on Build GRCh38

View Full Product Details