Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33488163-33488392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC28
Entrez Gene ID
54943
Gene Name
DnaJ heat shock protein family (Hsp40) member C28
Gene Aliases
C21orf55, C21orf78
UniGene
Hs.458308
Gene Chromosome Location
Chr.21: 33488055-33491723 on Build GRCh38
Transcripts
NM_001040192.2 NM_001320746.1 NM_017833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7365098 c.1033A>T p.I345L Chr.21: 33488361-33488361 on Build GRCh38
3550354 c.1008A>C p.R336S Chr.21: 33488386-33488386 on Build GRCh38
3972748 c.1130T>A p.M377K Chr.21: 33488264-33488264 on Build GRCh38
1666188 c.1073T>G p.L358R Chr.21: 33488321-33488321 on Build GRCh38
419254 c.1110G>C p.K370N Chr.21: 33488284-33488284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546367322 Chr.21: 33488237-33488237 on Build GRCh38
rs548149621 Chr.21: 33488307-33488309 on Build GRCh38
rs745998189 Chr.21: 33488339-33488339 on Build GRCh38
rs775370742 Chr.21: 33488345-33488345 on Build GRCh38
rs779358335 Chr.21: 33488314-33488314 on Build GRCh38
rs953496920 Chr.21: 33488303-33488303 on Build GRCh38
rs774449561 Chr.21: 33488216-33488216 on Build GRCh38
rs200537192 Chr.21: 33488279-33488279 on Build GRCh38
rs757788305 Chr.21: 33488307-33488307 on Build GRCh38
rs751212470 Chr.21: 33488285-33488285 on Build GRCh38
rs373310996 Chr.21: 33488357-33488357 on Build GRCh38
rs375527360 Chr.21: 33488275-33488275 on Build GRCh38
rs1019340762 Chr.21: 33488349-33488349 on Build GRCh38
rs775774536 Chr.21: 33488238-33488238 on Build GRCh38
rs534560693 Chr.21: 33488200-33488200 on Build GRCh38
rs763805342 Chr.21: 33488248-33488248 on Build GRCh38
rs771365218 Chr.21: 33488203-33488203 on Build GRCh38
rs748768239 Chr.21: 33488272-33488273 on Build GRCh38
rs772175920 Chr.21: 33488342-33488342 on Build GRCh38
rs778117320 Chr.21: 33488302-33488302 on Build GRCh38
rs1010543640 Chr.21: 33488254-33488254 on Build GRCh38
rs372061825 Chr.21: 33488274-33488274 on Build GRCh38
rs749616309 Chr.21: 33488304-33488304 on Build GRCh38
rs552657108 Chr.21: 33488262-33488262 on Build GRCh38
rs372123876 Chr.21: 33488257-33488257 on Build GRCh38
rs760442498 Chr.21: 33488239-33488239 on Build GRCh38
rs763992633 Chr.21: 33488251-33488251 on Build GRCh38
rs747258791 Chr.21: 33488354-33488354 on Build GRCh38
rs761873408 Chr.21: 33488261-33488261 on Build GRCh38
rs745934033 Chr.21: 33488219-33488219 on Build GRCh38
rs776588830 Chr.21: 33488366-33488366 on Build GRCh38
rs754570640 Chr.21: 33488286-33488286 on Build GRCh38

View Full Product Details