Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35475017-35475284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEAD3
Entrez Gene ID
7005
Gene Name
TEA domain transcription factor 3
Gene Aliases
DTEF-1, ETFR-1, TEAD-3, TEAD5, TEF-5, TEF5
UniGene
Hs.485205
Gene Chromosome Location
Chr.6: 35473597-35497084 on Build GRCh38
Transcripts
NM_003214.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5512247 c.1210G>A p.D404N Chr.6: 35475142-35475142 on Build GRCh38
1228850 c.1249G>A p.E417K Chr.6: 35475103-35475103 on Build GRCh38
1444014 c.1314G>A p.T438T Chr.6: 35475149-35475149 on Build GRCh38
5512249 c.1202C>T p.T401M Chr.6: 35475150-35475150 on Build GRCh38
1444013 c.1203G>A p.T401T Chr.6: 35475149-35475149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375296105 Chr.6: 35475182-35475182 on Build GRCh38
rs749509853 Chr.6: 35475149-35475149 on Build GRCh38
rs770710659 Chr.6: 35475162-35475162 on Build GRCh38
rs1042290123 Chr.6: 35475038-35475038 on Build GRCh38
rs760435941 Chr.6: 35475071-35475071 on Build GRCh38
rs773735537 Chr.6: 35475161-35475161 on Build GRCh38
rs755075136 Chr.6: 35475150-35475150 on Build GRCh38
rs1028318634 Chr.6: 35475165-35475165 on Build GRCh38
rs980693701 Chr.6: 35475242-35475242 on Build GRCh38
rs879549399 Chr.6: 35475212-35475214 on Build GRCh38
rs368857310 Chr.6: 35475128-35475128 on Build GRCh38
rs893226490 Chr.6: 35475154-35475154 on Build GRCh38
rs572348553 Chr.6: 35475255-35475255 on Build GRCh38
rs372235163 Chr.6: 35475144-35475144 on Build GRCh38
rs572216404 Chr.6: 35475259-35475259 on Build GRCh38
rs542976494 Chr.6: 35475265-35475265 on Build GRCh38
rs780189681 Chr.6: 35475145-35475145 on Build GRCh38
rs556619552 Chr.6: 35475079-35475079 on Build GRCh38
rs1008924763 Chr.6: 35475209-35475209 on Build GRCh38
rs776491907 Chr.6: 35475163-35475163 on Build GRCh38
rs891108815 Chr.6: 35475208-35475208 on Build GRCh38
rs773682085 Chr.6: 35475055-35475055 on Build GRCh38
rs752634565 Chr.6: 35475098-35475098 on Build GRCh38
rs538849907 Chr.6: 35475166-35475166 on Build GRCh38
rs201810762 Chr.6: 35475160-35475160 on Build GRCh38
rs758902471 Chr.6: 35475082-35475082 on Build GRCh38
rs751311912 Chr.6: 35475131-35475131 on Build GRCh38
rs775030252 Chr.6: 35475189-35475189 on Build GRCh38
rs41270072 Chr.6: 35475220-35475220 on Build GRCh38
rs961903562 Chr.6: 35475225-35475225 on Build GRCh38
rs374769894 Chr.6: 35475104-35475104 on Build GRCh38
rs759213534 Chr.6: 35475164-35475164 on Build GRCh38
rs772374927 Chr.6: 35475043-35475043 on Build GRCh38
rs868310029 Chr.6: 35475115-35475115 on Build GRCh38
rs371647902 Chr.6: 35475083-35475083 on Build GRCh38
rs1039475558 Chr.6: 35475077-35475077 on Build GRCh38

View Full Product Details