Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109463201-109463474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC24B
Entrez Gene ID
10427
Gene Name
SEC24 homolog B, COPII coat complex component
Gene Aliases
SEC24
UniGene
Hs.292472
Gene Chromosome Location
Chr.4: 109433715-109540459 on Build GRCh38
Transcripts
NM_001042734.3 NM_001300813.2 NM_001318085.1 NM_001318086.1 NM_006323.4 XM_005262688.1 XM_005262691.1 XM_011531535.2 XM_011531536.2 XM_011531537.2 XM_011531539.2 XM_017007652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1720366 c.529_530CC>TT p.P177F Chr.4: 109463296-109463297 on Build GRCh38
1753482 c.438G>T p.T146T Chr.4: 109463205-109463205 on Build GRCh38
3599009 c.650C>A p.P217Q Chr.4: 109463417-109463417 on Build GRCh38
447212 c.601C>T p.L201L Chr.4: 109463368-109463368 on Build GRCh38
5466015 c.677C>A p.S226* Chr.4: 109463444-109463444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770210135 Chr.4: 109463246-109463246 on Build GRCh38
rs752689419 Chr.4: 109463431-109463431 on Build GRCh38
rs766164314 Chr.4: 109463415-109463415 on Build GRCh38
rs566527942 Chr.4: 109463278-109463278 on Build GRCh38
rs148637147 Chr.4: 109463361-109463361 on Build GRCh38
rs759984821 Chr.4: 109463263-109463263 on Build GRCh38
rs376483793 Chr.4: 109463347-109463347 on Build GRCh38
rs781175458 Chr.4: 109463240-109463240 on Build GRCh38
rs778022531 Chr.4: 109463440-109463440 on Build GRCh38
rs538971019 Chr.4: 109463309-109463309 on Build GRCh38
rs377519783 Chr.4: 109463340-109463340 on Build GRCh38
rs756227690 Chr.4: 109463435-109463435 on Build GRCh38
rs370352426 Chr.4: 109463418-109463418 on Build GRCh38
rs374657201 Chr.4: 109463252-109463252 on Build GRCh38
rs751488571 Chr.4: 109463317-109463317 on Build GRCh38
rs767303675 Chr.4: 109463428-109463428 on Build GRCh38
rs370513339 Chr.4: 109463360-109463360 on Build GRCh38
rs530184881 Chr.4: 109463239-109463239 on Build GRCh38
rs201445712 Chr.4: 109463341-109463341 on Build GRCh38
rs781330260 Chr.4: 109463324-109463324 on Build GRCh38
rs983661457 Chr.4: 109463226-109463226 on Build GRCh38
rs758144411 Chr.4: 109463302-109463302 on Build GRCh38
rs199632271 Chr.4: 109463366-109463366 on Build GRCh38
rs367617863 Chr.4: 109463383-109463383 on Build GRCh38
rs999275489 Chr.4: 109463251-109463251 on Build GRCh38
rs748298454 Chr.4: 109463326-109463326 on Build GRCh38
rs368947555 Chr.4: 109463256-109463256 on Build GRCh38
rs779320948 Chr.4: 109463450-109463450 on Build GRCh38
rs746946726 Chr.4: 109463225-109463225 on Build GRCh38
rs750080147 Chr.4: 109463288-109463288 on Build GRCh38
rs773702405 Chr.4: 109463247-109463247 on Build GRCh38
rs373344282 Chr.4: 109463268-109463268 on Build GRCh38
rs558585475 Chr.4: 109463321-109463321 on Build GRCh38
rs754091537 Chr.4: 109463442-109463442 on Build GRCh38
rs769324853 Chr.4: 109463378-109463378 on Build GRCh38
rs761440185 Chr.4: 109463282-109463282 on Build GRCh38
rs748347278 Chr.4: 109463243-109463243 on Build GRCh38
rs558781877 Chr.4: 109463237-109463237 on Build GRCh38
rs751374672 Chr.4: 109463417-109463417 on Build GRCh38
rs373705741 Chr.4: 109463343-109463343 on Build GRCh38
rs775988787 Chr.4: 109463365-109463365 on Build GRCh38
rs922615041 Chr.4: 109463434-109463434 on Build GRCh38
rs764811622 Chr.4: 109463283-109463283 on Build GRCh38
rs201240080 Chr.4: 109463342-109463342 on Build GRCh38
rs762559535 Chr.4: 109463414-109463414 on Build GRCh38
rs942226097 Chr.4: 109463296-109463296 on Build GRCh38
rs778254272 Chr.4: 109463391-109463391 on Build GRCh38
rs200924216 Chr.4: 109463447-109463447 on Build GRCh38

View Full Product Details