Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20197382-20197619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR11G2
Entrez Gene ID
390439
Gene Name
olfactory receptor family 11 subfamily G member 2
Gene Aliases
OR14-34
UniGene
Hs.554582
Gene Chromosome Location
Chr.14: 20197336-20198373 on Build GRCh38
Transcripts
NM_001005503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6309226 c.134G>T p.S45I Chr.14: 20197469-20197469 on Build GRCh38
4470582 c.166C>T p.P56S Chr.14: 20197501-20197501 on Build GRCh38
6194796 c.146G>C p.G49A Chr.14: 20197481-20197481 on Build GRCh38
5499899 c.153C>G p.I51M Chr.14: 20197488-20197488 on Build GRCh38
6140077 c.97A>T p.R33W Chr.14: 20197432-20197432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009121030 Chr.14: 20197532-20197532 on Build GRCh38
rs779976178 Chr.14: 20197568-20197568 on Build GRCh38
rs758063205 Chr.14: 20197463-20197463 on Build GRCh38
rs768775067 Chr.14: 20197550-20197550 on Build GRCh38
rs532455728 Chr.14: 20197531-20197531 on Build GRCh38
rs766776011 Chr.14: 20197415-20197415 on Build GRCh38
rs748411393 Chr.14: 20197584-20197584 on Build GRCh38
rs749706829 Chr.14: 20197593-20197593 on Build GRCh38
rs778782038 Chr.14: 20197438-20197438 on Build GRCh38
rs965019685 Chr.14: 20197556-20197556 on Build GRCh38
rs1028111176 Chr.14: 20197573-20197573 on Build GRCh38
rs774718962 Chr.14: 20197524-20197524 on Build GRCh38
rs565585281 Chr.14: 20197430-20197430 on Build GRCh38
rs773370429 Chr.14: 20197590-20197590 on Build GRCh38
rs989748155 Chr.14: 20197601-20197601 on Build GRCh38
rs45620333 Chr.14: 20197468-20197468 on Build GRCh38
rs148888019 Chr.14: 20197426-20197426 on Build GRCh38
rs952559248 Chr.14: 20197575-20197575 on Build GRCh38
rs764808971 Chr.14: 20197558-20197558 on Build GRCh38
rs145505238 Chr.14: 20197418-20197418 on Build GRCh38
rs753394210 Chr.14: 20197436-20197436 on Build GRCh38
rs756663982 Chr.14: 20197551-20197551 on Build GRCh38
rs140668031 Chr.14: 20197481-20197481 on Build GRCh38
rs774386316 Chr.14: 20197592-20197592 on Build GRCh38
rs763281429 Chr.14: 20197412-20197412 on Build GRCh38
rs752277871 Chr.14: 20197416-20197416 on Build GRCh38
rs375394817 Chr.14: 20197521-20197521 on Build GRCh38
rs369399690 Chr.14: 20197564-20197564 on Build GRCh38
rs755251338 Chr.14: 20197576-20197576 on Build GRCh38
rs748310237 Chr.14: 20197497-20197497 on Build GRCh38
rs770325674 Chr.14: 20197501-20197501 on Build GRCh38
rs996875254 Chr.14: 20197557-20197557 on Build GRCh38
rs146843651 Chr.14: 20197473-20197473 on Build GRCh38
rs753341187 Chr.14: 20197549-20197549 on Build GRCh38
rs776823464 Chr.14: 20197490-20197490 on Build GRCh38
rs906612495 Chr.14: 20197535-20197535 on Build GRCh38
rs145451340 Chr.14: 20197427-20197427 on Build GRCh38
rs758385135 Chr.14: 20197566-20197566 on Build GRCh38
rs746842958 Chr.14: 20197570-20197570 on Build GRCh38
rs760296116 Chr.14: 20197525-20197525 on Build GRCh38
rs757188359 Chr.14: 20197437-20197437 on Build GRCh38
rs138827087 Chr.14: 20197518-20197518 on Build GRCh38
rs145848913 Chr.14: 20197586-20197586 on Build GRCh38
rs1035984368 Chr.14: 20197453-20197453 on Build GRCh38
rs1003985065 Chr.14: 20197544-20197544 on Build GRCh38
rs866049933 Chr.14: 20197506-20197506 on Build GRCh38
rs773529501 Chr.14: 20197514-20197514 on Build GRCh38
rs373432254 Chr.14: 20197454-20197454 on Build GRCh38

View Full Product Details