Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 14824153-14824426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
-
UniGene
Hs.439199
Gene Chromosome Location
Chr.Y: 14522578-14844945 on Build GRCh38
Transcripts
NM_001206850.1 NM_014893.4 NR_028319.1 NR_046355.1 XM_006724874.2 XM_011531424.2 XM_011531425.2 XM_011531426.2 XM_011531427.1 XM_011531428.1 XM_011531429.2 XM_011531430.2 XM_017030034.1 XM_017030035.1 XM_017030036.1 XM_017030037.1 XM_017030038.1 XM_017030039.1 XM_017030040.1

SNP Detail

SNP ID Chromosome Location
rs752615966 Chr.Y: 14824205-14824205 on Build GRCh38
rs756094037 Chr.Y: 14824400-14824400 on Build GRCh38
rs777521989 Chr.Y: 14824255-14824255 on Build GRCh38
rs368891956 Chr.Y: 14824267-14824267 on Build GRCh38
rs35394227 Chr.Y: 14824201-14824201 on Build GRCh38
rs758333597 Chr.Y: 14824214-14824214 on Build GRCh38
rs372147378 Chr.Y: 14824276-14824276 on Build GRCh38
rs866193447 Chr.Y: 14824355-14824355 on Build GRCh38
rs780170994 Chr.Y: 14824197-14824197 on Build GRCh38
rs774470645 Chr.Y: 14824390-14824390 on Build GRCh38
rs749495741 Chr.Y: 14824321-14824321 on Build GRCh38
rs752868113 Chr.Y: 14824295-14824295 on Build GRCh38
rs754596054 Chr.Y: 14824185-14824185 on Build GRCh38
rs768661445 Chr.Y: 14824336-14824336 on Build GRCh38

View Full Product Details