Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 118884638-118884889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX15
Entrez Gene ID
6913
Gene Name
T-box 15
Gene Aliases
TBX14
UniGene
Hs.146196
Gene Chromosome Location
Chr.1: 118883043-118989556 on Build GRCh38
Transcripts
NM_152380.2 XM_005271161.3 XM_005271162.1 XM_011542052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797876 c.1352A>G p.Y451C Chr.1: 118884871-118884871 on Build GRCh38
5377385 c.896C>T p.P299L Chr.1: 118884829-118884829 on Build GRCh38
6459189 c.1662G>T p.E554D Chr.1: 118884879-118884879 on Build GRCh38
6797875 c.940C>T p.Q314* Chr.1: 118884785-118884785 on Build GRCh38
4142249 c.1379T>G p.M460R Chr.1: 118884844-118884844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769321316 Chr.1: 118884783-118884783 on Build GRCh38
rs758186213 Chr.1: 118884739-118884739 on Build GRCh38
rs760984537 Chr.1: 118884711-118884711 on Build GRCh38
rs750744766 Chr.1: 118884710-118884710 on Build GRCh38
rs752659709 Chr.1: 118884725-118884725 on Build GRCh38
rs142137699 Chr.1: 118884850-118884850 on Build GRCh38
rs940938093 Chr.1: 118884763-118884763 on Build GRCh38
rs532340026 Chr.1: 118884693-118884693 on Build GRCh38
rs975217822 Chr.1: 118884713-118884713 on Build GRCh38
rs566094968 Chr.1: 118884792-118884792 on Build GRCh38
rs755047289 Chr.1: 118884717-118884717 on Build GRCh38
rs548905715 Chr.1: 118884820-118884820 on Build GRCh38
rs779624363 Chr.1: 118884869-118884869 on Build GRCh38
rs761076205 Chr.1: 118884822-118884822 on Build GRCh38
rs776131865 Chr.1: 118884690-118884690 on Build GRCh38
rs745406485 Chr.1: 118884778-118884778 on Build GRCh38
rs759037360 Chr.1: 118884691-118884691 on Build GRCh38
rs987600948 Chr.1: 118884760-118884760 on Build GRCh38
rs777452049 Chr.1: 118884684-118884684 on Build GRCh38
rs765347273 Chr.1: 118884845-118884845 on Build GRCh38
rs757011720 Chr.1: 118884859-118884859 on Build GRCh38
rs778757194 Chr.1: 118884722-118884722 on Build GRCh38
rs563818141 Chr.1: 118884788-118884788 on Build GRCh38
rs898141991 Chr.1: 118884775-118884775 on Build GRCh38
rs922050997 Chr.1: 118884746-118884746 on Build GRCh38
rs966627256 Chr.1: 118884682-118884682 on Build GRCh38
rs61730011 Chr.1: 118884844-118884844 on Build GRCh38
rs565063152 Chr.1: 118884670-118884670 on Build GRCh38
rs766655791 Chr.1: 118884714-118884714 on Build GRCh38
rs745518737 Chr.1: 118884866-118884866 on Build GRCh38
rs780781721 Chr.1: 118884766-118884766 on Build GRCh38
rs751385809 Chr.1: 118884858-118884858 on Build GRCh38
rs372032554 Chr.1: 118884718-118884718 on Build GRCh38
rs991778161 Chr.1: 118884666-118884666 on Build GRCh38
rs1037956589 Chr.1: 118884765-118884765 on Build GRCh38
rs948025394 Chr.1: 118884707-118884707 on Build GRCh38
rs770592627 Chr.1: 118884754-118884754 on Build GRCh38
rs902006435 Chr.1: 118884800-118884800 on Build GRCh38
rs12569041 Chr.1: 118884831-118884831 on Build GRCh38
rs375062885 Chr.1: 118884748-118884748 on Build GRCh38
rs762130621 Chr.1: 118884709-118884709 on Build GRCh38
rs916591526 Chr.1: 118884677-118884677 on Build GRCh38
rs182443245 Chr.1: 118884715-118884715 on Build GRCh38
rs938654317 Chr.1: 118884661-118884661 on Build GRCh38
rs752651011 Chr.1: 118884847-118884847 on Build GRCh38
rs940764815 Chr.1: 118884747-118884747 on Build GRCh38
rs150366466 Chr.1: 118884864-118884864 on Build GRCh38
rs771871257 Chr.1: 118884681-118884681 on Build GRCh38
rs200564235 Chr.1: 118884813-118884813 on Build GRCh38
rs755643362 Chr.1: 118884867-118884867 on Build GRCh38
rs868069252 Chr.1: 118884824-118884824 on Build GRCh38
rs887884962 Chr.1: 118884729-118884729 on Build GRCh38
rs763970919 Chr.1: 118884851-118884851 on Build GRCh38
rs908099540 Chr.1: 118884762-118884762 on Build GRCh38

View Full Product Details