Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103587227-103587488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOPT1
Entrez Gene ID
84334
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
APOP, APOP1, C14orf153
UniGene
Hs.598441
Gene Chromosome Location
Chr.14: 103562957-103592187 on Build GRCh38
Transcripts
NM_001302652.1 NM_001302653.1 NM_032374.4 NR_126431.1 NR_126432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6849009 c.437T>C p.F146S Chr.14: 103587325-103587325 on Build GRCh38
3700826 c.450T>A p.N150K Chr.14: 103587338-103587338 on Build GRCh38
6602482 c.502A>G p.M168V Chr.14: 103587351-103587351 on Build GRCh38
5428066 c.419C>T p.A140V Chr.14: 103587307-103587307 on Build GRCh38
6602486 c.420G>A p.A140A Chr.14: 103587308-103587308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373908486 Chr.14: 103587384-103587384 on Build GRCh38
rs745452081 Chr.14: 103587413-103587413 on Build GRCh38
rs947367355 Chr.14: 103587377-103587377 on Build GRCh38
rs776809478 Chr.14: 103587349-103587349 on Build GRCh38
rs761843894 Chr.14: 103587331-103587331 on Build GRCh38
rs149706370 Chr.14: 103587415-103587415 on Build GRCh38
rs942223588 Chr.14: 103587312-103587312 on Build GRCh38
rs935242172 Chr.14: 103587356-103587356 on Build GRCh38
rs759527186 Chr.14: 103587351-103587351 on Build GRCh38
rs745349308 Chr.14: 103587311-103587311 on Build GRCh38
rs771300575 Chr.14: 103587308-103587308 on Build GRCh38
rs2403197 Chr.14: 103587427-103587427 on Build GRCh38
rs114895750 Chr.14: 103587371-103587371 on Build GRCh38
rs780774719 Chr.14: 103587309-103587309 on Build GRCh38
rs781521385 Chr.14: 103587408-103587408 on Build GRCh38
rs900628637 Chr.14: 103587365-103587365 on Build GRCh38
rs755253849 Chr.14: 103587287-103587287 on Build GRCh38
rs750489685 Chr.14: 103587357-103587358 on Build GRCh38
rs751820338 Chr.14: 103587396-103587396 on Build GRCh38
rs182242924 Chr.14: 103587383-103587383 on Build GRCh38
rs562472810 Chr.14: 103587321-103587321 on Build GRCh38
rs368148821 Chr.14: 103587395-103587395 on Build GRCh38
rs779482530 Chr.14: 103587416-103587416 on Build GRCh38
rs765608908 Chr.14: 103587360-103587360 on Build GRCh38
rs758818892 Chr.14: 103587297-103587297 on Build GRCh38
rs747999315 Chr.14: 103587260-103587260 on Build GRCh38
rs931962880 Chr.14: 103587367-103587367 on Build GRCh38
rs753824369 Chr.14: 103587276-103587276 on Build GRCh38
rs757830782 Chr.14: 103587405-103587405 on Build GRCh38
rs145119626 Chr.14: 103587292-103587292 on Build GRCh38
rs778222675 Chr.14: 103587302-103587302 on Build GRCh38
rs747556529 Chr.14: 103587307-103587307 on Build GRCh38
rs547186048 Chr.14: 103587455-103587455 on Build GRCh38
rs774069678 Chr.14: 103587329-103587329 on Build GRCh38
rs147564236 Chr.14: 103587320-103587320 on Build GRCh38
rs1052785319 Chr.14: 103587358-103587358 on Build GRCh38
rs368317042 Chr.14: 103587333-103587333 on Build GRCh38
rs139609851 Chr.14: 103587288-103587288 on Build GRCh38
rs769478281 Chr.14: 103587319-103587319 on Build GRCh38

View Full Product Details