Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42685294-42685537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP1
Entrez Gene ID
8506
Gene Name
contactin associated protein 1
Gene Aliases
CASPR, CNTNAP, NRXN4, P190
UniGene
Hs.408730
Gene Chromosome Location
Chr.17: 42679963-42699993 on Build GRCh38
Transcripts
NM_003632.2 XM_005257748.4 XM_017025238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
979575 c.666C>T p.Y222Y Chr.17: 42685371-42685371 on Build GRCh38
979574 c.592G>A p.V198M Chr.17: 42685297-42685297 on Build GRCh38
460327 c.646G>A p.E216K Chr.17: 42685351-42685351 on Build GRCh38
5004919 c.645C>T p.A215A Chr.17: 42685350-42685350 on Build GRCh38
5825252 c.652G>A p.A218T Chr.17: 42685357-42685357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768387220 Chr.17: 42685347-42685347 on Build GRCh38
rs546838489 Chr.17: 42685464-42685465 on Build GRCh38
rs971683099 Chr.17: 42685441-42685441 on Build GRCh38
rs948394739 Chr.17: 42685449-42685449 on Build GRCh38
rs761624632 Chr.17: 42685449-42685450 on Build GRCh38
rs753641708 Chr.17: 42685398-42685398 on Build GRCh38
rs751029282 Chr.17: 42685447-42685447 on Build GRCh38
rs199605282 Chr.17: 42685432-42685432 on Build GRCh38
rs960306831 Chr.17: 42685477-42685477 on Build GRCh38
rs60104774 Chr.17: 42685501-42685501 on Build GRCh38
rs761328167 Chr.17: 42685366-42685366 on Build GRCh38
rs529351386 Chr.17: 42685460-42685460 on Build GRCh38
rs778544132 Chr.17: 42685461-42685461 on Build GRCh38
rs754704710 Chr.17: 42685377-42685377 on Build GRCh38
rs781415725 Chr.17: 42685440-42685440 on Build GRCh38
rs768847403 Chr.17: 42685457-42685457 on Build GRCh38
rs770121100 Chr.17: 42685335-42685335 on Build GRCh38
rs1052794932 Chr.17: 42685336-42685336 on Build GRCh38
rs748941568 Chr.17: 42685345-42685345 on Build GRCh38
rs773203084 Chr.17: 42685380-42685380 on Build GRCh38
rs114045118 Chr.17: 42685513-42685513 on Build GRCh38
rs547038534 Chr.17: 42685390-42685390 on Build GRCh38
rs778521443 Chr.17: 42685428-42685428 on Build GRCh38
rs371406732 Chr.17: 42685372-42685372 on Build GRCh38
rs749687032 Chr.17: 42685456-42685456 on Build GRCh38
rs368581393 Chr.17: 42685371-42685371 on Build GRCh38
rs759207925 Chr.17: 42685401-42685401 on Build GRCh38
rs747797746 Chr.17: 42685463-42685463 on Build GRCh38
rs563501193 Chr.17: 42685429-42685429 on Build GRCh38
rs1052851077 Chr.17: 42685516-42685516 on Build GRCh38
rs780385946 Chr.17: 42685455-42685455 on Build GRCh38
rs892358714 Chr.17: 42685348-42685348 on Build GRCh38
rs781767249 Chr.17: 42685311-42685311 on Build GRCh38
rs373521234 Chr.17: 42685450-42685450 on Build GRCh38
rs757416273 Chr.17: 42685438-42685438 on Build GRCh38
rs760650248 Chr.17: 42685382-42685382 on Build GRCh38
rs371727880 Chr.17: 42685436-42685436 on Build GRCh38
rs746309332 Chr.17: 42685333-42685333 on Build GRCh38
rs775909719 Chr.17: 42685339-42685339 on Build GRCh38

View Full Product Details