Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 126556413-126556670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM84B
Entrez Gene ID
157638
Gene Name
family with sequence similarity 84 member B
Gene Aliases
BCMP101, NSE2
UniGene
Hs.741352
Gene Chromosome Location
Chr.8: 126552438-126558499 on Build GRCh38
Transcripts
NM_174911.4 XM_017013107.1 XM_017013108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6478948 c.829G>T p.E277* Chr.8: 126556561-126556561 on Build GRCh38
7225859 c.823C>G p.P275A Chr.8: 126556567-126556567 on Build GRCh38
5512542 c.909_911delGGA p.E303delE Chr.8: 126556479-126556481 on Build GRCh38
1454780 c.812C>T p.T271M Chr.8: 126556578-126556578 on Build GRCh38
6530532 c.911A>G p.D304G Chr.8: 126556479-126556479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760250251 Chr.8: 126556494-126556494 on Build GRCh38
rs750491887 Chr.8: 126556531-126556531 on Build GRCh38
rs371552027 Chr.8: 126556490-126556490 on Build GRCh38
rs565358693 Chr.8: 126556508-126556508 on Build GRCh38
rs751633609 Chr.8: 126556462-126556462 on Build GRCh38
rs374699943 Chr.8: 126556546-126556546 on Build GRCh38
rs770726841 Chr.8: 126556551-126556551 on Build GRCh38
rs765132100 Chr.8: 126556506-126556506 on Build GRCh38
rs765038660 Chr.8: 126556449-126556449 on Build GRCh38
rs368094139 Chr.8: 126556582-126556582 on Build GRCh38
rs1056682555 Chr.8: 126556636-126556636 on Build GRCh38
rs767547351 Chr.8: 126556525-126556525 on Build GRCh38
rs779367719 Chr.8: 126556598-126556598 on Build GRCh38
rs576789380 Chr.8: 126556567-126556567 on Build GRCh38
rs778008176 Chr.8: 126556459-126556459 on Build GRCh38
rs144431781 Chr.8: 126556588-126556588 on Build GRCh38
rs774161569 Chr.8: 126556577-126556577 on Build GRCh38
rs748568137 Chr.8: 126556610-126556610 on Build GRCh38
rs750486835 Chr.8: 126556585-126556585 on Build GRCh38
rs774284867 Chr.8: 126556493-126556493 on Build GRCh38
rs937897160 Chr.8: 126556615-126556615 on Build GRCh38
rs371909525 Chr.8: 126556642-126556642 on Build GRCh38
rs776180702 Chr.8: 126556554-126556554 on Build GRCh38
rs759408368 Chr.8: 126556434-126556434 on Build GRCh38
rs898960203 Chr.8: 126556644-126556644 on Build GRCh38
rs567614477 Chr.8: 126556562-126556562 on Build GRCh38
rs746320331 Chr.8: 126556487-126556487 on Build GRCh38
rs375199157 Chr.8: 126556559-126556559 on Build GRCh38
rs757617035 Chr.8: 126556502-126556503 on Build GRCh38
rs746597425 Chr.8: 126556548-126556548 on Build GRCh38
rs960904623 Chr.8: 126556466-126556466 on Build GRCh38
rs376896029 Chr.8: 126556453-126556453 on Build GRCh38
rs780772163 Chr.8: 126556619-126556619 on Build GRCh38
rs754246135 Chr.8: 126556590-126556590 on Build GRCh38
rs778043627 Chr.8: 126556575-126556575 on Build GRCh38
rs941132916 Chr.8: 126556602-126556602 on Build GRCh38
rs752454857 Chr.8: 126556450-126556450 on Build GRCh38
rs755550800 Chr.8: 126556594-126556594 on Build GRCh38
rs751721294 Chr.8: 126556516-126556516 on Build GRCh38
rs79248636 Chr.8: 126556557-126556557 on Build GRCh38
rs757385407 Chr.8: 126556479-126556479 on Build GRCh38
rs545145275 Chr.8: 126556504-126556504 on Build GRCh38
rs915830243 Chr.8: 126556604-126556604 on Build GRCh38
rs764286582 Chr.8: 126556515-126556515 on Build GRCh38
rs531850133 Chr.8: 126556491-126556491 on Build GRCh38
rs749662617 Chr.8: 126556538-126556538 on Build GRCh38
rs770510445 Chr.8: 126556498-126556498 on Build GRCh38
rs770237284 Chr.8: 126556489-126556489 on Build GRCh38
rs367700277 Chr.8: 126556547-126556547 on Build GRCh38
rs1002534720 Chr.8: 126556441-126556441 on Build GRCh38
rs775346007 Chr.8: 126556566-126556566 on Build GRCh38
rs562691402 Chr.8: 126556486-126556486 on Build GRCh38
rs560326320 Chr.8: 126556521-126556521 on Build GRCh38
rs745369982 Chr.8: 126556635-126556635 on Build GRCh38
rs756624733 Chr.8: 126556532-126556532 on Build GRCh38
rs148176598 Chr.8: 126556612-126556612 on Build GRCh38

View Full Product Details