Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42159916-42160189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKH
Entrez Gene ID
160851
Gene Name
diacylglycerol kinase eta
Gene Aliases
DGKeta
UniGene
Hs.630380
Gene Chromosome Location
Chr.13: 42040036-42256584 on Build GRCh38
Transcripts
NM_001204504.2 NM_001204505.2 NM_001204506.2 NM_001297429.1 NM_152910.5 NM_178009.4 NR_123714.1 NR_123715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6998383 c.350G>T p.G117V Chr.13: 42160039-42160039 on Build GRCh38
256754 c.818G>A p.R273H Chr.13: 42160099-42160099 on Build GRCh38
6643014 c.326C>T p.A109V Chr.13: 42160015-42160015 on Build GRCh38
7364238 c.787G>C p.V263L Chr.13: 42160068-42160068 on Build GRCh38
947511 c.792C>T p.C264C Chr.13: 42160073-42160073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772276834 Chr.13: 42159977-42159977 on Build GRCh38
rs370149879 Chr.13: 42160099-42160099 on Build GRCh38
rs763818773 Chr.13: 42160036-42160036 on Build GRCh38
rs1051271492 Chr.13: 42160046-42160046 on Build GRCh38
rs757309834 Chr.13: 42160098-42160098 on Build GRCh38
rs370894389 Chr.13: 42160052-42160052 on Build GRCh38
rs775996653 Chr.13: 42160031-42160031 on Build GRCh38
rs138595528 Chr.13: 42160129-42160129 on Build GRCh38
rs1010911860 Chr.13: 42160120-42160120 on Build GRCh38
rs534054205 Chr.13: 42160042-42160042 on Build GRCh38
rs143249155 Chr.13: 42160068-42160068 on Build GRCh38
rs117428221 Chr.13: 42159971-42159971 on Build GRCh38
rs9562382 Chr.13: 42160040-42160040 on Build GRCh38
rs772464156 Chr.13: 42160133-42160133 on Build GRCh38
rs1041307243 Chr.13: 42159953-42159953 on Build GRCh38
rs770414433 Chr.13: 42160114-42160114 on Build GRCh38
rs147508042 Chr.13: 42160073-42160073 on Build GRCh38
rs770859410 Chr.13: 42160126-42160127 on Build GRCh38
rs769176952 Chr.13: 42160134-42160134 on Build GRCh38
rs376451530 Chr.13: 42159996-42159996 on Build GRCh38
rs756049566 Chr.13: 42160043-42160043 on Build GRCh38
rs1015182713 Chr.13: 42160044-42160044 on Build GRCh38
rs776191177 Chr.13: 42160119-42160119 on Build GRCh38
rs746031340 Chr.13: 42160109-42160109 on Build GRCh38
rs374871339 Chr.13: 42160106-42160106 on Build GRCh38
rs766369723 Chr.13: 42160047-42160047 on Build GRCh38
rs773005970 Chr.13: 42160019-42160020 on Build GRCh38
rs74051557 Chr.13: 42159993-42159993 on Build GRCh38
rs867182332 Chr.13: 42159949-42159949 on Build GRCh38
rs754814160 Chr.13: 42160058-42160058 on Build GRCh38
rs770222067 Chr.13: 42160025-42160025 on Build GRCh38
rs552462527 Chr.13: 42159950-42159950 on Build GRCh38
rs761386468 Chr.13: 42160155-42160155 on Build GRCh38
rs200670152 Chr.13: 42160035-42160035 on Build GRCh38
rs748487719 Chr.13: 42160132-42160132 on Build GRCh38
rs770954723 Chr.13: 42160014-42160014 on Build GRCh38
rs774007287 Chr.13: 42160135-42160135 on Build GRCh38
rs746380479 Chr.13: 42160016-42160016 on Build GRCh38
rs970979330 Chr.13: 42160066-42160066 on Build GRCh38
rs941262242 Chr.13: 42159957-42159957 on Build GRCh38
rs201484878 Chr.13: 42159970-42159970 on Build GRCh38
rs762675726 Chr.13: 42160023-42160024 on Build GRCh38
rs145824794 Chr.13: 42160015-42160015 on Build GRCh38

View Full Product Details