Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 54940088-54940349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83B
Entrez Gene ID
222584
Gene Name
family with sequence similarity 83 member B
Gene Aliases
C6orf143
UniGene
Hs.152423
Gene Chromosome Location
Chr.6: 54846323-54945099 on Build GRCh38
Transcripts
NM_001010872.2 XM_006715022.3 XM_011514394.2 XM_011514395.2 XM_017010478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718822 c.1199T>G p.L400R Chr.6: 54940170-54940170 on Build GRCh38
5720686 c.1286C>T p.S429L Chr.6: 54940257-54940257 on Build GRCh38
3830802 c.1285T>C p.S429P Chr.6: 54940256-54940256 on Build GRCh38
4818953 c.1271G>T p.S424I Chr.6: 54940242-54940242 on Build GRCh38
3830801 c.1236A>C p.P412P Chr.6: 54940207-54940207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029413066 Chr.6: 54940174-54940174 on Build GRCh38
rs114519346 Chr.6: 54940144-54940144 on Build GRCh38
rs776399930 Chr.6: 54940137-54940137 on Build GRCh38
rs752327982 Chr.6: 54940308-54940308 on Build GRCh38
rs370955123 Chr.6: 54940279-54940279 on Build GRCh38
rs769196988 Chr.6: 54940238-54940238 on Build GRCh38
rs866283097 Chr.6: 54940115-54940115 on Build GRCh38
rs149822989 Chr.6: 54940192-54940192 on Build GRCh38
rs767890059 Chr.6: 54940180-54940180 on Build GRCh38
rs13211183 Chr.6: 54940200-54940200 on Build GRCh38
rs1047039605 Chr.6: 54940303-54940303 on Build GRCh38
rs774667525 Chr.6: 54940165-54940165 on Build GRCh38
rs771043633 Chr.6: 54940249-54940249 on Build GRCh38
rs9475076 Chr.6: 54940276-54940276 on Build GRCh38
rs754623876 Chr.6: 54940228-54940228 on Build GRCh38
rs200139084 Chr.6: 54940173-54940173 on Build GRCh38
rs373963057 Chr.6: 54940160-54940160 on Build GRCh38
rs374175673 Chr.6: 54940239-54940239 on Build GRCh38
rs758777129 Chr.6: 54940295-54940295 on Build GRCh38
rs919519373 Chr.6: 54940269-54940269 on Build GRCh38
rs775741755 Chr.6: 54940321-54940321 on Build GRCh38
rs867463762 Chr.6: 54940145-54940145 on Build GRCh38
rs548232784 Chr.6: 54940255-54940255 on Build GRCh38
rs765802493 Chr.6: 54940281-54940281 on Build GRCh38
rs746277812 Chr.6: 54940222-54940222 on Build GRCh38
rs747640441 Chr.6: 54940237-54940237 on Build GRCh38
rs375673525 Chr.6: 54940194-54940194 on Build GRCh38
rs780131232 Chr.6: 54940215-54940216 on Build GRCh38
rs754301491 Chr.6: 54940216-54940216 on Build GRCh38
rs28592584 Chr.6: 54940149-54940149 on Build GRCh38
rs770123761 Chr.6: 54940211-54940212 on Build GRCh38
rs757574046 Chr.6: 54940218-54940218 on Build GRCh38
rs750774204 Chr.6: 54940292-54940292 on Build GRCh38
rs372267421 Chr.6: 54940248-54940248 on Build GRCh38
rs147775708 Chr.6: 54940317-54940317 on Build GRCh38
rs780830259 Chr.6: 54940297-54940297 on Build GRCh38
rs760741144 Chr.6: 54940268-54940268 on Build GRCh38
rs769761541 Chr.6: 54940143-54940143 on Build GRCh38
rs141100648 Chr.6: 54940318-54940318 on Build GRCh38
rs996582264 Chr.6: 54940132-54940132 on Build GRCh38
rs759812852 Chr.6: 54940168-54940168 on Build GRCh38
rs779268613 Chr.6: 54940219-54940219 on Build GRCh38
rs746097903 Chr.6: 54940210-54940211 on Build GRCh38
rs748850090 Chr.6: 54940315-54940315 on Build GRCh38
rs776090893 Chr.6: 54940130-54940130 on Build GRCh38
rs553145456 Chr.6: 54940310-54940310 on Build GRCh38
rs573151626 Chr.6: 54940311-54940311 on Build GRCh38
rs866880366 Chr.6: 54940312-54940312 on Build GRCh38
rs866900742 Chr.6: 54940140-54940140 on Build GRCh38
rs376048871 Chr.6: 54940161-54940161 on Build GRCh38
rs149060290 Chr.6: 54940259-54940259 on Build GRCh38
rs775587388 Chr.6: 54940267-54940267 on Build GRCh38
rs771764348 Chr.6: 54940320-54940320 on Build GRCh38
rs568083070 Chr.6: 54940270-54940270 on Build GRCh38

View Full Product Details