Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30158646-30158875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL5
Entrez Gene ID
51646
Gene Name
yippee like 5
Gene Aliases
CGI-127
UniGene
Hs.515890
Gene Chromosome Location
Chr.2: 30146568-30160533 on Build GRCh38
Transcripts
NM_001127399.1 NM_001127400.1 NM_001127401.1 NM_016061.2 XM_017004318.1 XM_017004319.1 XM_017004320.1 XM_017004321.1 XM_017004322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5455072 c.202G>A p.V68I Chr.2: 30158679-30158679 on Build GRCh38
1668829 c.268G>T p.D90Y Chr.2: 30158745-30158745 on Build GRCh38
7182934 c.295G>A p.V99M Chr.2: 30158772-30158772 on Build GRCh38
721275 c.307C>T p.R103C Chr.2: 30158784-30158784 on Build GRCh38
404705 c.346G>A p.V116I Chr.2: 30158823-30158823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756253472 Chr.2: 30158848-30158848 on Build GRCh38
rs750711475 Chr.2: 30158804-30158804 on Build GRCh38
rs760185286 Chr.2: 30158771-30158771 on Build GRCh38
rs748427434 Chr.2: 30158842-30158842 on Build GRCh38
rs765654576 Chr.2: 30158789-30158789 on Build GRCh38
rs758802312 Chr.2: 30158821-30158821 on Build GRCh38
rs960842189 Chr.2: 30158796-30158796 on Build GRCh38
rs772836658 Chr.2: 30158666-30158666 on Build GRCh38
rs749256772 Chr.2: 30158681-30158681 on Build GRCh38
rs534446426 Chr.2: 30158759-30158759 on Build GRCh38
rs752096275 Chr.2: 30158826-30158826 on Build GRCh38
rs200449455 Chr.2: 30158707-30158707 on Build GRCh38
rs755116618 Chr.2: 30158837-30158837 on Build GRCh38
rs759106199 Chr.2: 30158755-30158755 on Build GRCh38
rs374303662 Chr.2: 30158823-30158823 on Build GRCh38
rs201738526 Chr.2: 30158762-30158762 on Build GRCh38
rs370225674 Chr.2: 30158769-30158769 on Build GRCh38
rs1025934749 Chr.2: 30158785-30158785 on Build GRCh38
rs528828815 Chr.2: 30158839-30158839 on Build GRCh38
rs201650008 Chr.2: 30158688-30158688 on Build GRCh38

View Full Product Details