Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28329099-28329346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNOC
Entrez Gene ID
5368
Gene Name
prepronociceptin
Gene Aliases
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ
UniGene
Hs.88218
Gene Chromosome Location
Chr.8: 28316979-28343355 on Build GRCh38
Transcripts
NM_006228.4 XM_005273532.2 XM_011544559.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6896441 c.39C>G p.L13L Chr.8: 28329196-28329196 on Build GRCh38
145787 c.37C>G p.L13V Chr.8: 28329194-28329194 on Build GRCh38
1098873 c.50T>C p.V17A Chr.8: 28329207-28329207 on Build GRCh38
6358714 c.124G>T p.E42* Chr.8: 28329281-28329281 on Build GRCh38
1569088 c.35G>A p.S12N Chr.8: 28329192-28329192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777196606 Chr.8: 28329283-28329283 on Build GRCh38
rs975295753 Chr.8: 28329254-28329254 on Build GRCh38
rs537112845 Chr.8: 28329320-28329320 on Build GRCh38
rs753974288 Chr.8: 28329266-28329266 on Build GRCh38
rs759886616 Chr.8: 28329247-28329247 on Build GRCh38
rs982660560 Chr.8: 28329118-28329118 on Build GRCh38
rs752580573 Chr.8: 28329280-28329280 on Build GRCh38
rs781084231 Chr.8: 28329277-28329277 on Build GRCh38
rs746221971 Chr.8: 28329318-28329318 on Build GRCh38
rs754926931 Chr.8: 28329174-28329174 on Build GRCh38
rs777286757 Chr.8: 28329221-28329221 on Build GRCh38
rs781054493 Chr.8: 28329199-28329199 on Build GRCh38
rs557567617 Chr.8: 28329281-28329281 on Build GRCh38
rs756930743 Chr.8: 28329313-28329313 on Build GRCh38
rs945099289 Chr.8: 28329302-28329302 on Build GRCh38
rs556788041 Chr.8: 28329321-28329321 on Build GRCh38
rs1051300399 Chr.8: 28329292-28329292 on Build GRCh38
rs751323221 Chr.8: 28329162-28329162 on Build GRCh38
rs776188984 Chr.8: 28329258-28329258 on Build GRCh38
rs904021845 Chr.8: 28329309-28329309 on Build GRCh38
rs199907549 Chr.8: 28329256-28329256 on Build GRCh38
rs962656620 Chr.8: 28329190-28329190 on Build GRCh38
rs778471219 Chr.8: 28329317-28329317 on Build GRCh38
rs1037509369 Chr.8: 28329271-28329271 on Build GRCh38
rs755112945 Chr.8: 28329314-28329316 on Build GRCh38
rs764233620 Chr.8: 28329261-28329261 on Build GRCh38
rs941905387 Chr.8: 28329154-28329154 on Build GRCh38
rs778806214 Chr.8: 28329314-28329315 on Build GRCh38
rs757588559 Chr.8: 28329121-28329121 on Build GRCh38
rs774766551 Chr.8: 28329244-28329244 on Build GRCh38
rs909714944 Chr.8: 28329144-28329144 on Build GRCh38
rs190892464 Chr.8: 28329275-28329275 on Build GRCh38
rs769343150 Chr.8: 28329216-28329216 on Build GRCh38
rs149853236 Chr.8: 28329312-28329313 on Build GRCh38
rs747622540 Chr.8: 28329210-28329210 on Build GRCh38
rs187303222 Chr.8: 28329237-28329237 on Build GRCh38
rs374745274 Chr.8: 28329270-28329270 on Build GRCh38
rs368188580 Chr.8: 28329274-28329274 on Build GRCh38
rs762034105 Chr.8: 28329269-28329269 on Build GRCh38
rs28415085 Chr.8: 28329304-28329304 on Build GRCh38
rs913678259 Chr.8: 28329293-28329293 on Build GRCh38
rs748898197 Chr.8: 28329227-28329227 on Build GRCh38
rs369595539 Chr.8: 28329155-28329155 on Build GRCh38
rs779835364 Chr.8: 28329159-28329159 on Build GRCh38

View Full Product Details