Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51944985-51945228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP3
Entrez Gene ID
10039
Gene Name
poly(ADP-ribose) polymerase family member 3
Gene Aliases
ADPRT3, ADPRTL2, ADPRTL3, ARTD3, IRT1, PADPRT-3
UniGene
Hs.271742
Gene Chromosome Location
Chr.3: 51942282-51948867 on Build GRCh38
Transcripts
NM_001003931.3 NM_005485.5 XM_005264779.4 XM_017005490.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
382282 c.737C>A p.A246D Chr.3: 51945079-51945079 on Build GRCh38
5708822 c.731C>T p.T244M Chr.3: 51945094-51945094 on Build GRCh38
5708821 c.752C>T p.T251M Chr.3: 51945094-51945094 on Build GRCh38
1424316 c.833C>T p.P278L Chr.3: 51945175-51945175 on Build GRCh38
6737904 c.830A>G p.E277G Chr.3: 51945193-51945193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771397204 Chr.3: 51945065-51945065 on Build GRCh38
rs764250768 Chr.3: 51945184-51945184 on Build GRCh38
rs777853119 Chr.3: 51945125-51945125 on Build GRCh38
rs367984364 Chr.3: 51945017-51945017 on Build GRCh38
rs199681575 Chr.3: 51945067-51945067 on Build GRCh38
rs79176238 Chr.3: 51945148-51945148 on Build GRCh38
rs781639763 Chr.3: 51945134-51945134 on Build GRCh38
rs759376439 Chr.3: 51945171-51945171 on Build GRCh38
rs761582982 Chr.3: 51945029-51945029 on Build GRCh38
rs767301379 Chr.3: 51945032-51945032 on Build GRCh38
rs995264319 Chr.3: 51945086-51945086 on Build GRCh38
rs761446569 Chr.3: 51945082-51945082 on Build GRCh38
rs1025275811 Chr.3: 51945087-51945087 on Build GRCh38
rs763703635 Chr.3: 51945023-51945023 on Build GRCh38
rs373954134 Chr.3: 51945203-51945203 on Build GRCh38
rs750079296 Chr.3: 51945054-51945062 on Build GRCh38
rs375042601 Chr.3: 51945140-51945140 on Build GRCh38
rs758563354 Chr.3: 51945114-51945114 on Build GRCh38
rs755984921 Chr.3: 51945037-51945037 on Build GRCh38
rs765147770 Chr.3: 51945174-51945174 on Build GRCh38
rs369155045 Chr.3: 51945141-51945141 on Build GRCh38
rs1009098407 Chr.3: 51945059-51945059 on Build GRCh38
rs760383165 Chr.3: 51945094-51945094 on Build GRCh38
rs200544465 Chr.3: 51945054-51945054 on Build GRCh38
rs774090571 Chr.3: 51945024-51945024 on Build GRCh38
rs371960861 Chr.3: 51945131-51945131 on Build GRCh38
rs752698881 Chr.3: 51945175-51945175 on Build GRCh38
rs761144798 Chr.3: 51945011-51945011 on Build GRCh38
rs35082474 Chr.3: 51945031-51945032 on Build GRCh38
rs367595307 Chr.3: 51945062-51945062 on Build GRCh38
rs747264733 Chr.3: 51945126-51945126 on Build GRCh38
rs750153146 Chr.3: 51945036-51945036 on Build GRCh38
rs6413415 Chr.3: 51945059-51945061 on Build GRCh38
rs778334099 Chr.3: 51945146-51945146 on Build GRCh38
rs766208894 Chr.3: 51945038-51945038 on Build GRCh38
rs762278473 Chr.3: 51945147-51945147 on Build GRCh38
rs754332150 Chr.3: 51945137-51945137 on Build GRCh38
rs767050625 Chr.3: 51945089-51945089 on Build GRCh38
rs750791962 Chr.3: 51945206-51945206 on Build GRCh38
rs375713100 Chr.3: 51945093-51945093 on Build GRCh38
rs912447475 Chr.3: 51945202-51945202 on Build GRCh38
rs774027071 Chr.3: 51945069-51945069 on Build GRCh38
rs762213299 Chr.3: 51945017-51945017 on Build GRCh38
rs323870 Chr.3: 51945169-51945169 on Build GRCh38
rs562584096 Chr.3: 51945176-51945176 on Build GRCh38
rs1051490353 Chr.3: 51945028-51945028 on Build GRCh38
rs751685874 Chr.3: 51945189-51945189 on Build GRCh38
rs753879087 Chr.3: 51945039-51945039 on Build GRCh38
rs765590755 Chr.3: 51945037-51945038 on Build GRCh38
rs771324673 Chr.3: 51945012-51945012 on Build GRCh38
rs375724604 Chr.3: 51945149-51945149 on Build GRCh38
rs540910959 Chr.3: 51945068-51945068 on Build GRCh38
rs4986825 Chr.3: 51945104-51945104 on Build GRCh38
rs777257572 Chr.3: 51945014-51945014 on Build GRCh38
rs564192189 Chr.3: 51945045-51945045 on Build GRCh38
rs760388246 Chr.3: 51945158-51945158 on Build GRCh38
rs762529302 Chr.3: 51945124-51945124 on Build GRCh38
rs149821166 Chr.3: 51945046-51945046 on Build GRCh38
rs753716622 Chr.3: 51945095-51945095 on Build GRCh38
rs770580933 Chr.3: 51945159-51945159 on Build GRCh38
rs986719196 Chr.3: 51945188-51945188 on Build GRCh38
rs752753892 Chr.3: 51945113-51945113 on Build GRCh38
rs756601875 Chr.3: 51945209-51945209 on Build GRCh38

View Full Product Details