Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69031423-69031684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOX5
Entrez Gene ID
79400
Gene Name
NADPH oxidase, EF-hand calcium binding domain 5
Gene Aliases
-
UniGene
Hs.657932
Gene Chromosome Location
Chr.15: 68930500-69057161 on Build GRCh38
Transcripts
NM_001184779.1 NM_001184780.1 NM_024505.3 NR_033671.2 NR_033672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6341180 c.319G>T p.A107S Chr.15: 69031565-69031565 on Build GRCh38
4643919 c.362G>T p.G121V Chr.15: 69031608-69031608 on Build GRCh38
5008384 c.427C>G p.P143A Chr.15: 69031619-69031619 on Build GRCh38
6376241 c.338A>G p.Q113R Chr.15: 69031530-69031530 on Build GRCh38
6341179 c.373G>T p.A125S Chr.15: 69031565-69031565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766750205 Chr.15: 69031501-69031501 on Build GRCh38
rs746671904 Chr.15: 69031589-69031589 on Build GRCh38
rs367806524 Chr.15: 69031554-69031554 on Build GRCh38
rs372991107 Chr.15: 69031631-69031631 on Build GRCh38
rs368928363 Chr.15: 69031570-69031570 on Build GRCh38
rs989280256 Chr.15: 69031494-69031494 on Build GRCh38
rs771463645 Chr.15: 69031600-69031600 on Build GRCh38
rs774813093 Chr.15: 69031615-69031615 on Build GRCh38
rs768203012 Chr.15: 69031487-69031487 on Build GRCh38
rs760836024 Chr.15: 69031452-69031452 on Build GRCh38
rs745697423 Chr.15: 69031549-69031549 on Build GRCh38
rs938340734 Chr.15: 69031450-69031450 on Build GRCh38
rs779833747 Chr.15: 69031633-69031633 on Build GRCh38
rs758338742 Chr.15: 69031632-69031632 on Build GRCh38
rs776154979 Chr.15: 69031598-69031598 on Build GRCh38
rs779633500 Chr.15: 69031577-69031577 on Build GRCh38
rs775578798 Chr.15: 69031620-69031620 on Build GRCh38
rs753733900 Chr.15: 69031538-69031538 on Build GRCh38
rs772334710 Chr.15: 69031664-69031664 on Build GRCh38
rs773662196 Chr.15: 69031493-69031493 on Build GRCh38
rs1012864310 Chr.15: 69031499-69031499 on Build GRCh38
rs141822986 Chr.15: 69031635-69031635 on Build GRCh38
rs751190593 Chr.15: 69031634-69031634 on Build GRCh38
rs368305444 Chr.15: 69031532-69031532 on Build GRCh38
rs747701698 Chr.15: 69031647-69031647 on Build GRCh38
rs535867207 Chr.15: 69031463-69031463 on Build GRCh38
rs565915241 Chr.15: 69031473-69031473 on Build GRCh38
rs780674027 Chr.15: 69031642-69031642 on Build GRCh38
rs549372850 Chr.15: 69031471-69031471 on Build GRCh38
rs145992353 Chr.15: 69031522-69031522 on Build GRCh38
rs758740642 Chr.15: 69031628-69031629 on Build GRCh38
rs757259289 Chr.15: 69031655-69031655 on Build GRCh38
rs753930782 Chr.15: 69031625-69031625 on Build GRCh38
rs943321593 Chr.15: 69031584-69031584 on Build GRCh38
rs746553979 Chr.15: 69031482-69031482 on Build GRCh38
rs769279947 Chr.15: 69031649-69031649 on Build GRCh38
rs200180547 Chr.15: 69031629-69031629 on Build GRCh38
rs757381672 Chr.15: 69031628-69031628 on Build GRCh38
rs763391449 Chr.15: 69031498-69031498 on Build GRCh38
rs764166594 Chr.15: 69031535-69031535 on Build GRCh38
rs764105375 Chr.15: 69031621-69031621 on Build GRCh38
rs78571685 Chr.15: 69031619-69031619 on Build GRCh38
rs774655468 Chr.15: 69031502-69031502 on Build GRCh38
rs752949926 Chr.15: 69031526-69031526 on Build GRCh38
rs779462144 Chr.15: 69031659-69031659 on Build GRCh38
rs767776363 Chr.15: 69031524-69031524 on Build GRCh38
rs757212780 Chr.15: 69031545-69031545 on Build GRCh38
rs746306193 Chr.15: 69031663-69031663 on Build GRCh38
rs768058504 Chr.15: 69031594-69031594 on Build GRCh38

View Full Product Details