Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41840101-41840362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD7-PLA2G4B
Entrez Gene ID
8681
Gene Name
JMJD7-PLA2G4B readthrough
Gene Aliases
HsT16992, cPLA2-beta
UniGene
Hs.198161
Gene Chromosome Location
Chr.15: 41828085-41848148 on Build GRCh38
Transcripts
NM_001198588.1 NM_005090.3 NM_001114633.1

Gene Symbol
PLA2G4B
Entrez Gene ID
100137049
Gene Name
phospholipase A2 group IVB
Gene Aliases
HsT16992, cPLA2-beta
UniGene
Hs.198161
Gene Chromosome Location
Chr.15: 41838813-41848148 on Build GRCh38
Transcripts
NM_001198588.1 NM_005090.3 NM_001114633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4054613 c.731C>T p.T244M Chr.15: 41840186-41840186 on Build GRCh38
4753670 c.754C>T p.R252C Chr.15: 41840209-41840209 on Build GRCh38
3816044 c.744G>A p.L248L Chr.15: 41840199-41840199 on Build GRCh38
2186464 c.708G>T p.E236D Chr.15: 41840163-41840163 on Build GRCh38
2186465 c.775+8C>T p.? Chr.15: 41840238-41840238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144829225 Chr.15: 41840221-41840221 on Build GRCh38
rs372691513 Chr.15: 41840276-41840276 on Build GRCh38
rs771566095 Chr.15: 41840161-41840161 on Build GRCh38
rs779580470 Chr.15: 41840270-41840270 on Build GRCh38
rs755979718 Chr.15: 41840150-41840150 on Build GRCh38
rs368218325 Chr.15: 41840239-41840239 on Build GRCh38
rs547983742 Chr.15: 41840270-41840284 on Build GRCh38
rs370285316 Chr.15: 41840126-41840126 on Build GRCh38
rs750430804 Chr.15: 41840268-41840268 on Build GRCh38
rs758741732 Chr.15: 41840279-41840281 on Build GRCh38
rs980855710 Chr.15: 41840324-41840324 on Build GRCh38
rs764501222 Chr.15: 41840168-41840168 on Build GRCh38
rs896925689 Chr.15: 41840212-41840212 on Build GRCh38
rs375174855 Chr.15: 41840219-41840219 on Build GRCh38
rs985643305 Chr.15: 41840325-41840325 on Build GRCh38
rs149781451 Chr.15: 41840186-41840186 on Build GRCh38
rs751885905 Chr.15: 41840192-41840192 on Build GRCh38
rs757826472 Chr.15: 41840269-41840269 on Build GRCh38
rs570415180 Chr.15: 41840154-41840154 on Build GRCh38
rs778923237 Chr.15: 41840211-41840211 on Build GRCh38
rs998798007 Chr.15: 41840327-41840327 on Build GRCh38
rs922590784 Chr.15: 41840320-41840320 on Build GRCh38
rs558348016 Chr.15: 41840279-41840279 on Build GRCh38
rs752341383 Chr.15: 41840153-41840153 on Build GRCh38
rs1008727178 Chr.15: 41840313-41840313 on Build GRCh38
rs768253127 Chr.15: 41840277-41840277 on Build GRCh38
rs941506936 Chr.15: 41840340-41840340 on Build GRCh38
rs976886748 Chr.15: 41840284-41840284 on Build GRCh38
rs888914989 Chr.15: 41840288-41840288 on Build GRCh38
rs1033952598 Chr.15: 41840332-41840332 on Build GRCh38
rs745989136 Chr.15: 41840230-41840230 on Build GRCh38
rs555635500 Chr.15: 41840238-41840238 on Build GRCh38
rs777837634 Chr.15: 41840152-41840152 on Build GRCh38
rs778910532 Chr.15: 41840140-41840140 on Build GRCh38
rs1010094236 Chr.15: 41840256-41840256 on Build GRCh38
rs775127588 Chr.15: 41840162-41840162 on Build GRCh38
rs750627912 Chr.15: 41840141-41840141 on Build GRCh38
rs777100888 Chr.15: 41840175-41840175 on Build GRCh38
rs143606352 Chr.15: 41840209-41840209 on Build GRCh38
rs745405146 Chr.15: 41840158-41840158 on Build GRCh38
rs761025591 Chr.15: 41840163-41840163 on Build GRCh38
rs764517193 Chr.15: 41840133-41840133 on Build GRCh38
rs780349655 Chr.15: 41840145-41840145 on Build GRCh38
rs765470111 Chr.15: 41840218-41840218 on Build GRCh38
rs368677050 Chr.15: 41840139-41840139 on Build GRCh38
rs754411016 Chr.15: 41840136-41840136 on Build GRCh38
rs750424910 Chr.15: 41840191-41840191 on Build GRCh38
rs763234818 Chr.15: 41840258-41840258 on Build GRCh38
rs762264503 Chr.15: 41840237-41840237 on Build GRCh38
rs375396549 Chr.15: 41840250-41840250 on Build GRCh38
rs369389884 Chr.15: 41840252-41840252 on Build GRCh38
rs148060081 Chr.15: 41840225-41840225 on Build GRCh38
rs539537890 Chr.15: 41840155-41840155 on Build GRCh38
rs922658486 Chr.15: 41840142-41840142 on Build GRCh38
rs765809431 Chr.15: 41840187-41840187 on Build GRCh38
rs778708576 Chr.15: 41840265-41840265 on Build GRCh38
rs547576405 Chr.15: 41840146-41840146 on Build GRCh38
rs372024053 Chr.15: 41840322-41840322 on Build GRCh38
rs568407095 Chr.15: 41840121-41840121 on Build GRCh38
rs75341740 Chr.15: 41840127-41840127 on Build GRCh38
rs569293395 Chr.15: 41840210-41840210 on Build GRCh38
rs543877319 Chr.15: 41840336-41840336 on Build GRCh38
rs755948695 Chr.15: 41840195-41840195 on Build GRCh38
rs372133894 Chr.15: 41840143-41840143 on Build GRCh38
rs910137624 Chr.15: 41840329-41840329 on Build GRCh38
rs201446325 Chr.15: 41840244-41840244 on Build GRCh38
rs757350087 Chr.15: 41840138-41840138 on Build GRCh38
rs115974862 Chr.15: 41840243-41840243 on Build GRCh38
rs776941260 Chr.15: 41840233-41840233 on Build GRCh38
rs578013087 Chr.15: 41840321-41840321 on Build GRCh38
rs773777176 Chr.15: 41840240-41840240 on Build GRCh38
rs757078475 Chr.15: 41840253-41840253 on Build GRCh38

View Full Product Details