Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176329250-176329475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTX2
Entrez Gene ID
10651
Gene Name
metaxin 2
Gene Aliases
-
UniGene
Hs.470728
Gene Chromosome Location
Chr.2: 176269395-176338025 on Build GRCh38
Transcripts
NM_001006635.2 NM_001319097.1 NM_001319098.1 NM_006554.4 XM_011510497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6043432 c.469C>T p.R157C Chr.2: 176329382-176329382 on Build GRCh38
5463048 c.483A>C p.K161N Chr.2: 176329366-176329366 on Build GRCh38
5000460 c.447C>G p.Y149* Chr.2: 176329330-176329330 on Build GRCh38
39382 c.515T>A p.I172N Chr.2: 176329398-176329398 on Build GRCh38
3961565 c.425A>G p.H142R Chr.2: 176329308-176329308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747412662 Chr.2: 176329310-176329310 on Build GRCh38
rs969077633 Chr.2: 176329326-176329326 on Build GRCh38
rs756942943 Chr.2: 176329430-176329430 on Build GRCh38
rs201522166 Chr.2: 176329359-176329359 on Build GRCh38
rs747391856 Chr.2: 176329444-176329444 on Build GRCh38
rs772281523 Chr.2: 176329276-176329276 on Build GRCh38
rs773540891 Chr.2: 176329354-176329354 on Build GRCh38
rs755589338 Chr.2: 176329406-176329406 on Build GRCh38
rs552510181 Chr.2: 176329363-176329363 on Build GRCh38
rs1021522899 Chr.2: 176329306-176329306 on Build GRCh38
rs777294999 Chr.2: 176329417-176329417 on Build GRCh38
rs752678493 Chr.2: 176329383-176329383 on Build GRCh38
rs780486769 Chr.2: 176329304-176329304 on Build GRCh38
rs141452692 Chr.2: 176329319-176329319 on Build GRCh38
rs566651317 Chr.2: 176329323-176329323 on Build GRCh38
rs949333174 Chr.2: 176329274-176329274 on Build GRCh38
rs769206893 Chr.2: 176329311-176329311 on Build GRCh38
rs770050494 Chr.2: 176329330-176329330 on Build GRCh38
rs200408701 Chr.2: 176329382-176329382 on Build GRCh38
rs750590378 Chr.2: 176329297-176329297 on Build GRCh38
rs763268568 Chr.2: 176329356-176329356 on Build GRCh38
rs777347290 Chr.2: 176329315-176329315 on Build GRCh38
rs774430488 Chr.2: 176329383-176329384 on Build GRCh38
rs370986623 Chr.2: 176329429-176329429 on Build GRCh38
rs780214191 Chr.2: 176329433-176329433 on Build GRCh38
rs375769448 Chr.2: 176329289-176329289 on Build GRCh38
rs759326434 Chr.2: 176329366-176329366 on Build GRCh38
rs748581416 Chr.2: 176329305-176329305 on Build GRCh38
rs199730223 Chr.2: 176329423-176329423 on Build GRCh38

View Full Product Details