Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220762847-220763120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARC2
Entrez Gene ID
54996
Gene Name
mitochondrial amidoxime reducing component 2
Gene Aliases
MOSC2
UniGene
Hs.369042
Gene Chromosome Location
Chr.1: 220748246-220784815 on Build GRCh38
Transcripts
NM_001317338.1 NM_017898.4 XM_006711407.2 XM_011509684.1 XM_017001581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1959338 c.649G>A p.D217N Chr.1: 220762949-220762949 on Build GRCh38
7156644 c.636C>G p.L212L Chr.1: 220762936-220762936 on Build GRCh38
3803954 c.667T>C p.L223L Chr.1: 220762967-220762967 on Build GRCh38
4902753 c.681G>A p.M227I Chr.1: 220762981-220762981 on Build GRCh38
3741019 c.627C>T p.Y209Y Chr.1: 220762927-220762927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377671353 Chr.1: 220762875-220762875 on Build GRCh38
rs751669650 Chr.1: 220762939-220762939 on Build GRCh38
rs866055510 Chr.1: 220762974-220762974 on Build GRCh38
rs202006158 Chr.1: 220762942-220762942 on Build GRCh38
rs756220352 Chr.1: 220762940-220762940 on Build GRCh38
rs137931375 Chr.1: 220762933-220762933 on Build GRCh38
rs541434400 Chr.1: 220763019-220763019 on Build GRCh38
rs777375018 Chr.1: 220763097-220763097 on Build GRCh38
rs753886516 Chr.1: 220763075-220763075 on Build GRCh38
rs772650406 Chr.1: 220762904-220762904 on Build GRCh38
rs752545149 Chr.1: 220762926-220762926 on Build GRCh38
rs752347965 Chr.1: 220763033-220763033 on Build GRCh38
rs746659210 Chr.1: 220762889-220762892 on Build GRCh38
rs376827992 Chr.1: 220763029-220763029 on Build GRCh38
rs563293765 Chr.1: 220763063-220763063 on Build GRCh38
rs149492776 Chr.1: 220762991-220762991 on Build GRCh38
rs754663530 Chr.1: 220762920-220762920 on Build GRCh38
rs760255555 Chr.1: 220762915-220762915 on Build GRCh38
rs76664695 Chr.1: 220763035-220763035 on Build GRCh38
rs780249694 Chr.1: 220763067-220763067 on Build GRCh38
rs374115952 Chr.1: 220762872-220762872 on Build GRCh38
rs1040816309 Chr.1: 220762899-220762899 on Build GRCh38
rs200282208 Chr.1: 220763094-220763094 on Build GRCh38
rs768864833 Chr.1: 220762957-220762957 on Build GRCh38
rs767848879 Chr.1: 220763040-220763040 on Build GRCh38
rs773232890 Chr.1: 220763023-220763023 on Build GRCh38
rs182872 Chr.1: 220763078-220763078 on Build GRCh38
rs756562298 Chr.1: 220763062-220763062 on Build GRCh38
rs768205761 Chr.1: 220763025-220763032 on Build GRCh38
rs371425032 Chr.1: 220762928-220762928 on Build GRCh38
rs899609790 Chr.1: 220762929-220762929 on Build GRCh38
rs555805753 Chr.1: 220762954-220762954 on Build GRCh38
rs3795535 Chr.1: 220763030-220763030 on Build GRCh38
rs756375791 Chr.1: 220763096-220763096 on Build GRCh38
rs775182486 Chr.1: 220762884-220762884 on Build GRCh38
rs760465034 Chr.1: 220763026-220763026 on Build GRCh38
rs774122543 Chr.1: 220762871-220762871 on Build GRCh38
rs764961352 Chr.1: 220763032-220763032 on Build GRCh38
rs770489495 Chr.1: 220763028-220763028 on Build GRCh38
rs747033810 Chr.1: 220763098-220763098 on Build GRCh38
rs767704285 Chr.1: 220763038-220763038 on Build GRCh38
rs190616095 Chr.1: 220763056-220763056 on Build GRCh38
rs932801684 Chr.1: 220762977-220762977 on Build GRCh38
rs753534521 Chr.1: 220762919-220762919 on Build GRCh38
rs765769266 Chr.1: 220762917-220762917 on Build GRCh38
rs868149005 Chr.1: 220763012-220763012 on Build GRCh38
rs776987579 Chr.1: 220763091-220763091 on Build GRCh38
rs757887186 Chr.1: 220762932-220762932 on Build GRCh38
rs777367156 Chr.1: 220763099-220763099 on Build GRCh38
rs943718978 Chr.1: 220762880-220762880 on Build GRCh38
rs778724274 Chr.1: 220762967-220762967 on Build GRCh38
rs201205507 Chr.1: 220763039-220763039 on Build GRCh38
rs779132115 Chr.1: 220763079-220763079 on Build GRCh38

View Full Product Details