Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102074221-102074456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP37
Entrez Gene ID
79023
Gene Name
nucleoporin 37
Gene Aliases
p37
UniGene
Hs.444276
Gene Chromosome Location
Chr.12: 102074189-102120139 on Build GRCh38
Transcripts
NM_024057.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
691600 c.935T>C p.V312A Chr.12: 102074400-102074400 on Build GRCh38
4512805 c.916C>T p.R306* Chr.12: 102074419-102074419 on Build GRCh38
3732381 c.969G>T p.V323V Chr.12: 102074366-102074366 on Build GRCh38
5431521 c.928C>T p.L310L Chr.12: 102074407-102074407 on Build GRCh38
934081 c.967delG p.V323fs*1 Chr.12: 102074368-102074368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038984444 Chr.12: 102074267-102074267 on Build GRCh38
rs761638403 Chr.12: 102074340-102074340 on Build GRCh38
rs771289715 Chr.12: 102074311-102074311 on Build GRCh38
rs150083754 Chr.12: 102074352-102074352 on Build GRCh38
rs775237165 Chr.12: 102074411-102074412 on Build GRCh38
rs373141402 Chr.12: 102074325-102074325 on Build GRCh38
rs186104441 Chr.12: 102074318-102074318 on Build GRCh38
rs777818201 Chr.12: 102074397-102074397 on Build GRCh38
rs758478568 Chr.12: 102074378-102074378 on Build GRCh38
rs1008872066 Chr.12: 102074252-102074252 on Build GRCh38
rs997184570 Chr.12: 102074288-102074288 on Build GRCh38
rs746341112 Chr.12: 102074419-102074419 on Build GRCh38
rs368671382 Chr.12: 102074418-102074418 on Build GRCh38
rs943279410 Chr.12: 102074256-102074256 on Build GRCh38
rs757604427 Chr.12: 102074407-102074407 on Build GRCh38
rs1028654798 Chr.12: 102074368-102074368 on Build GRCh38
rs767605689 Chr.12: 102074379-102074379 on Build GRCh38
rs374470945 Chr.12: 102074387-102074387 on Build GRCh38
rs763601948 Chr.12: 102074328-102074328 on Build GRCh38
rs756240742 Chr.12: 102074388-102074388 on Build GRCh38
rs760091797 Chr.12: 102074321-102074321 on Build GRCh38
rs901477341 Chr.12: 102074283-102074283 on Build GRCh38
rs572857209 Chr.12: 102074303-102074303 on Build GRCh38
rs867293542 Chr.12: 102074434-102074434 on Build GRCh38
rs753554593 Chr.12: 102074330-102074330 on Build GRCh38
rs760349784 Chr.12: 102074327-102074327 on Build GRCh38
rs771563760 Chr.12: 102074323-102074323 on Build GRCh38
rs545339061 Chr.12: 102074308-102074308 on Build GRCh38
rs749524333 Chr.12: 102074403-102074403 on Build GRCh38
rs765141248 Chr.12: 102074341-102074341 on Build GRCh38

View Full Product Details