Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 71134501-71134726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPHOSPH10
Entrez Gene ID
10199
Gene Name
M-phase phosphoprotein 10
Gene Aliases
CT90, MPP10, MPP10P, PPP1R106
UniGene
Hs.656208
Gene Chromosome Location
Chr.2: 71130314-71150102 on Build GRCh38
Transcripts
NM_005791.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022596 c.941A>G p.D314G Chr.2: 71134640-71134640 on Build GRCh38
5798876 c.958G>C p.D320H Chr.2: 71134657-71134657 on Build GRCh38
1022597 c.996C>A p.T332T Chr.2: 71134695-71134695 on Build GRCh38
1022598 c.1016C>T p.A339V Chr.2: 71134715-71134715 on Build GRCh38
721944 c.1007C>A p.P336Q Chr.2: 71134706-71134706 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753716098 Chr.2: 71134666-71134666 on Build GRCh38
rs1018844895 Chr.2: 71134559-71134559 on Build GRCh38
rs752125493 Chr.2: 71134646-71134646 on Build GRCh38
rs777031056 Chr.2: 71134585-71134585 on Build GRCh38
rs530817475 Chr.2: 71134550-71134550 on Build GRCh38
rs773267079 Chr.2: 71134639-71134639 on Build GRCh38
rs756554887 Chr.2: 71134594-71134594 on Build GRCh38
rs1006992107 Chr.2: 71134529-71134529 on Build GRCh38
rs147873518 Chr.2: 71134694-71134694 on Build GRCh38
rs746062664 Chr.2: 71134592-71134592 on Build GRCh38
rs766242051 Chr.2: 71134651-71134651 on Build GRCh38
rs377211972 Chr.2: 71134622-71134622 on Build GRCh38
rs140795709 Chr.2: 71134677-71134677 on Build GRCh38
rs775386676 Chr.2: 71134619-71134619 on Build GRCh38
rs764144694 Chr.2: 71134632-71134632 on Build GRCh38
rs760740373 Chr.2: 71134642-71134642 on Build GRCh38
rs770090784 Chr.2: 71134604-71134604 on Build GRCh38
rs759507287 Chr.2: 71134565-71134565 on Build GRCh38
rs1040523984 Chr.2: 71134532-71134532 on Build GRCh38

View Full Product Details