Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28197144-28197395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAN3
Entrez Gene ID
255967
Gene Name
PAN3 poly(A) specific ribonuclease subunit
Gene Aliases
-
UniGene
Hs.645015
Gene Chromosome Location
Chr.13: 28138079-28295338 on Build GRCh38
Transcripts
NM_175854.7 XM_005266332.3 XM_005266333.3 XM_005266334.1 XM_011535032.2 XM_011535033.2 XM_011535034.2 XM_017020528.1 XM_017020529.1 XM_017020530.1 XM_017020531.1 XM_017020532.1 XM_017020533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1366207 c.815T>C p.M272T Chr.13: 28197309-28197309 on Build GRCh38
7121169 c.710G>T p.G237V Chr.13: 28197204-28197204 on Build GRCh38
2070197 c.704G>A p.R235H Chr.13: 28197198-28197198 on Build GRCh38
6315673 c.776G>T p.C259F Chr.13: 28197270-28197270 on Build GRCh38
1659058 c.751C>T p.R251* Chr.13: 28197245-28197245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774711821 Chr.13: 28197201-28197201 on Build GRCh38
rs762350801 Chr.13: 28197174-28197175 on Build GRCh38
rs530757846 Chr.13: 28197178-28197178 on Build GRCh38
rs894250575 Chr.13: 28197332-28197332 on Build GRCh38
rs989918874 Chr.13: 28197231-28197231 on Build GRCh38
rs754036440 Chr.13: 28197228-28197228 on Build GRCh38
rs199583702 Chr.13: 28197241-28197241 on Build GRCh38
rs767743339 Chr.13: 28197204-28197204 on Build GRCh38
rs757082745 Chr.13: 28197323-28197323 on Build GRCh38
rs773655572 Chr.13: 28197283-28197283 on Build GRCh38
rs759729387 Chr.13: 28197319-28197319 on Build GRCh38
rs765282758 Chr.13: 28197238-28197240 on Build GRCh38
rs765187373 Chr.13: 28197321-28197321 on Build GRCh38
rs774962706 Chr.13: 28197270-28197270 on Build GRCh38
rs755094232 Chr.13: 28197229-28197229 on Build GRCh38
rs374150030 Chr.13: 28197302-28197302 on Build GRCh38
rs907238087 Chr.13: 28197263-28197263 on Build GRCh38
rs760932779 Chr.13: 28197216-28197216 on Build GRCh38
rs766569962 Chr.13: 28197223-28197223 on Build GRCh38
rs772510364 Chr.13: 28197276-28197276 on Build GRCh38
rs948506253 Chr.13: 28197339-28197339 on Build GRCh38
rs754089470 Chr.13: 28197310-28197310 on Build GRCh38
rs763458596 Chr.13: 28197190-28197190 on Build GRCh38
rs376399343 Chr.13: 28197172-28197172 on Build GRCh38
rs751535247 Chr.13: 28197322-28197322 on Build GRCh38
rs748859113 Chr.13: 28197260-28197260 on Build GRCh38
rs745480151 Chr.13: 28197173-28197173 on Build GRCh38
rs747972690 Chr.13: 28197251-28197251 on Build GRCh38
rs750690809 Chr.13: 28197206-28197206 on Build GRCh38
rs1046222455 Chr.13: 28197245-28197245 on Build GRCh38
rs768033902 Chr.13: 28197265-28197265 on Build GRCh38
rs370785599 Chr.13: 28197230-28197230 on Build GRCh38
rs750288545 Chr.13: 28197352-28197352 on Build GRCh38
rs769222561 Chr.13: 28197193-28197193 on Build GRCh38
rs769312693 Chr.13: 28197176-28197176 on Build GRCh38
rs780798629 Chr.13: 28197246-28197246 on Build GRCh38
rs377685978 Chr.13: 28197330-28197330 on Build GRCh38
rs761114999 Chr.13: 28197301-28197301 on Build GRCh38
rs779679483 Chr.13: 28197253-28197253 on Build GRCh38
rs762287056 Chr.13: 28197202-28197202 on Build GRCh38
rs1055299963 Chr.13: 28197317-28197317 on Build GRCh38
rs762187636 Chr.13: 28197271-28197271 on Build GRCh38

View Full Product Details