Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47009343-47009616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRF1
Entrez Gene ID
266977
Gene Name
adhesion G protein-coupled receptor F1
Gene Aliases
GPR110, KPG_012, PGR19, hGPCR36
UniGene
Hs.733762
Gene Chromosome Location
Chr.6: 46997703-47042363 on Build GRCh38
Transcripts
NM_153840.3 XM_011514469.1 XM_017010735.1 XM_017010736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3353362 c.2081C>A p.A694D Chr.6: 47009354-47009354 on Build GRCh38
5870838 c.1870C>T p.R624C Chr.6: 47009565-47009565 on Build GRCh38
7369886 c.1483C>A p.H495N Chr.6: 47009361-47009361 on Build GRCh38
1079974 c.1868G>A p.R623H Chr.6: 47009567-47009567 on Build GRCh38
3353363 c.1917C>G p.V639V Chr.6: 47009518-47009518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367711868 Chr.6: 47009573-47009573 on Build GRCh38
rs1011717035 Chr.6: 47009495-47009495 on Build GRCh38
rs149889820 Chr.6: 47009510-47009510 on Build GRCh38
rs572686616 Chr.6: 47009396-47009396 on Build GRCh38
rs761163287 Chr.6: 47009574-47009574 on Build GRCh38
rs745392518 Chr.6: 47009419-47009419 on Build GRCh38
rs779910886 Chr.6: 47009447-47009447 on Build GRCh38
rs142453870 Chr.6: 47009506-47009506 on Build GRCh38
rs747979631 Chr.6: 47009549-47009549 on Build GRCh38
rs753240558 Chr.6: 47009381-47009381 on Build GRCh38
rs140268316 Chr.6: 47009498-47009498 on Build GRCh38
rs772726495 Chr.6: 47009433-47009434 on Build GRCh38
rs759766802 Chr.6: 47009464-47009464 on Build GRCh38
rs139711395 Chr.6: 47009371-47009371 on Build GRCh38
rs759717417 Chr.6: 47009569-47009569 on Build GRCh38
rs151335777 Chr.6: 47009568-47009568 on Build GRCh38
rs372272153 Chr.6: 47009452-47009452 on Build GRCh38
rs907497974 Chr.6: 47009459-47009459 on Build GRCh38
rs776690262 Chr.6: 47009457-47009457 on Build GRCh38
rs773097753 Chr.6: 47009485-47009485 on Build GRCh38
rs763044746 Chr.6: 47009364-47009364 on Build GRCh38
rs983616269 Chr.6: 47009440-47009440 on Build GRCh38
rs766570587 Chr.6: 47009486-47009486 on Build GRCh38
rs781659590 Chr.6: 47009376-47009376 on Build GRCh38
rs143178573 Chr.6: 47009564-47009564 on Build GRCh38
rs765941297 Chr.6: 47009586-47009586 on Build GRCh38
rs144891661 Chr.6: 47009521-47009521 on Build GRCh38
rs760997937 Chr.6: 47009585-47009585 on Build GRCh38
rs138895375 Chr.6: 47009565-47009565 on Build GRCh38
rs767733749 Chr.6: 47009570-47009570 on Build GRCh38
rs144374951 Chr.6: 47009462-47009462 on Build GRCh38
rs933727564 Chr.6: 47009423-47009424 on Build GRCh38
rs544506995 Chr.6: 47009496-47009496 on Build GRCh38
rs768613174 Chr.6: 47009456-47009456 on Build GRCh38
rs16875406 Chr.6: 47009541-47009541 on Build GRCh38
rs200517235 Chr.6: 47009518-47009518 on Build GRCh38
rs138431849 Chr.6: 47009581-47009581 on Build GRCh38
rs183498225 Chr.6: 47009465-47009465 on Build GRCh38
rs769800523 Chr.6: 47009472-47009472 on Build GRCh38
rs772760006 Chr.6: 47009388-47009388 on Build GRCh38
rs374843683 Chr.6: 47009562-47009562 on Build GRCh38
rs766628746 Chr.6: 47009370-47009370 on Build GRCh38
rs140960279 Chr.6: 47009395-47009395 on Build GRCh38
rs764504592 Chr.6: 47009579-47009579 on Build GRCh38
rs753139873 Chr.6: 47009507-47009507 on Build GRCh38
rs749887956 Chr.6: 47009519-47009519 on Build GRCh38
rs771570431 Chr.6: 47009426-47009426 on Build GRCh38
rs992912537 Chr.6: 47009555-47009555 on Build GRCh38
rs757994346 Chr.6: 47009520-47009520 on Build GRCh38
rs146696622 Chr.6: 47009489-47009489 on Build GRCh38
rs746709993 Chr.6: 47009539-47009539 on Build GRCh38
rs141336103 Chr.6: 47009567-47009567 on Build GRCh38
rs368585261 Chr.6: 47009382-47009382 on Build GRCh38
rs773668060 Chr.6: 47009568-47009568 on Build GRCh38

View Full Product Details