Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11200566-11200795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XKR6
Entrez Gene ID
286046
Gene Name
XK related 6
Gene Aliases
C8orf21, C8orf5, C8orf7, XRG6
UniGene
Hs.657083
Gene Chromosome Location
Chr.8: 10896144-11205789 on Build GRCh38
Transcripts
NM_173683.3 XM_011543821.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6823833 c.552C>T p.F184F Chr.8: 11200788-11200788 on Build GRCh38
5962154 c.690G>A p.A230A Chr.8: 11200650-11200650 on Build GRCh38
6823848 c.688delG p.A230fs*51 Chr.8: 11200652-11200652 on Build GRCh38
6823835 c.699G>A p.L233L Chr.8: 11200641-11200641 on Build GRCh38
4162594 c.716G>T p.W239L Chr.8: 11200624-11200624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759138465 Chr.8: 11200721-11200721 on Build GRCh38
rs746222610 Chr.8: 11200651-11200651 on Build GRCh38
rs756407457 Chr.8: 11200762-11200762 on Build GRCh38
rs781533689 Chr.8: 11200746-11200746 on Build GRCh38
rs752115227 Chr.8: 11200727-11200727 on Build GRCh38
rs747499308 Chr.8: 11200635-11200635 on Build GRCh38
rs1036752440 Chr.8: 11200741-11200741 on Build GRCh38
rs750165179 Chr.8: 11200596-11200596 on Build GRCh38
rs766126309 Chr.8: 11200599-11200599 on Build GRCh38
rs766132838 Chr.8: 11200712-11200712 on Build GRCh38
rs753410459 Chr.8: 11200611-11200611 on Build GRCh38
rs903740691 Chr.8: 11200731-11200731 on Build GRCh38
rs753499996 Chr.8: 11200713-11200713 on Build GRCh38
rs369443880 Chr.8: 11200773-11200773 on Build GRCh38
rs771874571 Chr.8: 11200700-11200700 on Build GRCh38
rs778172698 Chr.8: 11200629-11200629 on Build GRCh38
rs755871497 Chr.8: 11200598-11200598 on Build GRCh38
rs1040111328 Chr.8: 11200657-11200657 on Build GRCh38
rs757679308 Chr.8: 11200641-11200641 on Build GRCh38
rs369205575 Chr.8: 11200694-11200694 on Build GRCh38
rs1045686094 Chr.8: 11200690-11200690 on Build GRCh38
rs1049656330 Chr.8: 11200587-11200587 on Build GRCh38
rs957339533 Chr.8: 11200653-11200653 on Build GRCh38
rs760488954 Chr.8: 11200702-11200702 on Build GRCh38
rs781647549 Chr.8: 11200643-11200643 on Build GRCh38
rs770063327 Chr.8: 11200659-11200659 on Build GRCh38
rs768684995 Chr.8: 11200666-11200666 on Build GRCh38
rs775635300 Chr.8: 11200660-11200660 on Build GRCh38
rs1042343137 Chr.8: 11200726-11200726 on Build GRCh38
rs924728890 Chr.8: 11200650-11200650 on Build GRCh38
rs774179481 Chr.8: 11200672-11200672 on Build GRCh38
rs754469665 Chr.8: 11200620-11200620 on Build GRCh38
rs757845278 Chr.8: 11200734-11200734 on Build GRCh38
rs749377142 Chr.8: 11200662-11200662 on Build GRCh38
rs553473562 Chr.8: 11200724-11200724 on Build GRCh38
rs901534540 Chr.8: 11200692-11200692 on Build GRCh38

View Full Product Details