Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 122152101-122152354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100505841
Entrez Gene ID
100505841
Gene Name
zinc finger protein 474-like
Gene Aliases
-
UniGene
Hs.733368
Gene Chromosome Location
Chr.5: 122129546-122182663 on Build GRCh38
Transcripts
XM_017008912.1 NM_207317.2

Gene Symbol
ZNF474
Entrez Gene ID
133923
Gene Name
zinc finger protein 474
Gene Aliases
-
UniGene
Hs.729673
Gene Chromosome Location
Chr.5: 122129513-122153571 on Build GRCh38
Transcripts
XM_017008912.1 NM_207317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3724294 c.184A>G p.R62G Chr.5: 122152174-122152174 on Build GRCh38
1581971 c.233C>T p.S78L Chr.5: 122152223-122152223 on Build GRCh38
3770308 c.256G>T p.V86L Chr.5: 122152246-122152246 on Build GRCh38
3724295 c.134C>T p.T45I Chr.5: 122152124-122152124 on Build GRCh38
6830512 c.234G>A p.S78S Chr.5: 122152224-122152224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774196594 Chr.5: 122152275-122152275 on Build GRCh38
rs771944641 Chr.5: 122152155-122152155 on Build GRCh38
rs751807350 Chr.5: 122152310-122152310 on Build GRCh38
rs114116635 Chr.5: 122152254-122152254 on Build GRCh38
rs76277653 Chr.5: 122152191-122152191 on Build GRCh38
rs949661912 Chr.5: 122152126-122152126 on Build GRCh38
rs114823456 Chr.5: 122152223-122152223 on Build GRCh38
rs758081957 Chr.5: 122152209-122152209 on Build GRCh38
rs116158782 Chr.5: 122152259-122152259 on Build GRCh38
rs377641114 Chr.5: 122152294-122152294 on Build GRCh38
rs776956245 Chr.5: 122152280-122152280 on Build GRCh38
rs72786994 Chr.5: 122152274-122152274 on Build GRCh38
rs765166888 Chr.5: 122152284-122152284 on Build GRCh38
rs761268305 Chr.5: 122152178-122152178 on Build GRCh38
rs571187077 Chr.5: 122152238-122152239 on Build GRCh38
rs759139646 Chr.5: 122152163-122152163 on Build GRCh38
rs772497468 Chr.5: 122152232-122152232 on Build GRCh38
rs777650163 Chr.5: 122152156-122152156 on Build GRCh38
rs535496939 Chr.5: 122152240-122152240 on Build GRCh38
rs747875906 Chr.5: 122152151-122152151 on Build GRCh38
rs150340655 Chr.5: 122152174-122152174 on Build GRCh38
rs762040729 Chr.5: 122152169-122152169 on Build GRCh38
rs905328107 Chr.5: 122152143-122152143 on Build GRCh38
rs760104498 Chr.5: 122152283-122152283 on Build GRCh38
rs111823798 Chr.5: 122152286-122152286 on Build GRCh38
rs144735279 Chr.5: 122152272-122152272 on Build GRCh38
rs937066114 Chr.5: 122152250-122152250 on Build GRCh38
rs143759385 Chr.5: 122152243-122152243 on Build GRCh38
rs374281616 Chr.5: 122152211-122152211 on Build GRCh38
rs1002376544 Chr.5: 122152165-122152165 on Build GRCh38
rs114373483 Chr.5: 122152276-122152276 on Build GRCh38
rs925148011 Chr.5: 122152247-122152247 on Build GRCh38
rs753954067 Chr.5: 122152182-122152182 on Build GRCh38
rs750259405 Chr.5: 122152328-122152328 on Build GRCh38
rs759496352 Chr.5: 122152245-122152245 on Build GRCh38
rs755150950 Chr.5: 122152130-122152130 on Build GRCh38
rs775639565 Chr.5: 122152292-122152292 on Build GRCh38
rs746974279 Chr.5: 122152157-122152157 on Build GRCh38
rs764265906 Chr.5: 122152295-122152295 on Build GRCh38
rs200772777 Chr.5: 122152320-122152320 on Build GRCh38
rs770373908 Chr.5: 122152159-122152159 on Build GRCh38
rs533290881 Chr.5: 122152304-122152304 on Build GRCh38
rs191501947 Chr.5: 122152224-122152224 on Build GRCh38
rs771454858 Chr.5: 122152278-122152278 on Build GRCh38
rs558433028 Chr.5: 122152258-122152258 on Build GRCh38
rs764372003 Chr.5: 122152256-122152256 on Build GRCh38
rs754407458 Chr.5: 122152221-122152221 on Build GRCh38
rs113839901 Chr.5: 122152128-122152128 on Build GRCh38
rs756072727 Chr.5: 122152330-122152330 on Build GRCh38
rs758655525 Chr.5: 122152255-122152255 on Build GRCh38
rs868184181 Chr.5: 122152138-122152138 on Build GRCh38
rs116012229 Chr.5: 122152273-122152273 on Build GRCh38
rs114918153 Chr.5: 122152185-122152185 on Build GRCh38
rs374646457 Chr.5: 122152315-122152315 on Build GRCh38
rs775244948 Chr.5: 122152167-122152167 on Build GRCh38
rs746884978 Chr.5: 122152218-122152218 on Build GRCh38
rs750900457 Chr.5: 122152177-122152177 on Build GRCh38
rs116318604 Chr.5: 122152266-122152266 on Build GRCh38
rs757185726 Chr.5: 122152219-122152219 on Build GRCh38
rs866996878 Chr.5: 122152172-122152172 on Build GRCh38
rs778973827 Chr.5: 122152186-122152186 on Build GRCh38
rs779253501 Chr.5: 122152131-122152131 on Build GRCh38
rs767059635 Chr.5: 122152179-122152179 on Build GRCh38
rs749154520 Chr.5: 122152141-122152141 on Build GRCh38
rs200600820 Chr.5: 122152160-122152160 on Build GRCh38
rs572551346 Chr.5: 122152253-122152253 on Build GRCh38

View Full Product Details