Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 38593675-38593904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18A
Entrez Gene ID
253650
Gene Name
ankyrin repeat domain 18A
Gene Aliases
-
UniGene
Hs.644630
Gene Chromosome Location
Chr.9: 38566260-38620587 on Build GRCh38
Transcripts
NM_147195.2 XM_006716749.3 XM_011517836.2 XM_011517837.2 XM_011517839.2 XM_017014576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
36859 c.1960G>A p.E654K Chr.9: 38593804-38593804 on Build GRCh38
4138953 c.1937C>T p.S646L Chr.9: 38593827-38593827 on Build GRCh38
6308134 c.1866A>T p.R622S Chr.9: 38593898-38593898 on Build GRCh38
1109134 c.1919C>A p.S640Y Chr.9: 38593845-38593845 on Build GRCh38
5823820 c.1974A>T p.S658S Chr.9: 38593790-38593790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773336450 Chr.9: 38593779-38593779 on Build GRCh38
rs967833881 Chr.9: 38593729-38593729 on Build GRCh38
rs267602244 Chr.9: 38593804-38593804 on Build GRCh38
rs975813538 Chr.9: 38593833-38593833 on Build GRCh38
rs964736468 Chr.9: 38593812-38593812 on Build GRCh38
rs535353908 Chr.9: 38593785-38593785 on Build GRCh38
rs1047985062 Chr.9: 38593869-38593869 on Build GRCh38
rs929567888 Chr.9: 38593865-38593865 on Build GRCh38
rs570939876 Chr.9: 38593734-38593734 on Build GRCh38
rs910270306 Chr.9: 38593777-38593777 on Build GRCh38
rs866964882 Chr.9: 38593838-38593838 on Build GRCh38
rs771467586 Chr.9: 38593860-38593860 on Build GRCh38
rs766542079 Chr.9: 38593852-38593852 on Build GRCh38
rs147995476 Chr.9: 38593823-38593823 on Build GRCh38

View Full Product Details