Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101488722-101488967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
UniGene
Hs.729062
Gene Chromosome Location
Chr.X: 101418263-101535988 on Build GRCh38
Transcripts
NM_001256155.2 NR_028407.2 NR_045861.1 NR_045862.1 NR_045863.1 NR_045864.1

SNP Detail

SNP ID Chromosome Location
rs5951332 Chr.X: 101488839-101488839 on Build GRCh38
rs782359403 Chr.X: 101488806-101488806 on Build GRCh38
rs1055839444 Chr.X: 101488825-101488825 on Build GRCh38
rs781974980 Chr.X: 101488794-101488794 on Build GRCh38
rs189267413 Chr.X: 101488774-101488774 on Build GRCh38
rs782795725 Chr.X: 101488848-101488848 on Build GRCh38
rs1020931861 Chr.X: 101488945-101488945 on Build GRCh38
rs782385528 Chr.X: 101488807-101488807 on Build GRCh38
rs782089176 Chr.X: 101488795-101488795 on Build GRCh38
rs1014378797 Chr.X: 101488934-101488934 on Build GRCh38
rs940661052 Chr.X: 101488754-101488754 on Build GRCh38

View Full Product Details