Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113739162-113739431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZW10
Entrez Gene ID
9183
Gene Name
zw10 kinetochore protein
Gene Aliases
HZW10, KNTC1AP
UniGene
Hs.503886
Gene Chromosome Location
Chr.11: 113733183-113773763 on Build GRCh38
Transcripts
NM_004724.3 XM_017018558.1 XM_017018559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
119027 c.1584-1G>C p.? Chr.11: 113739383-113739383 on Build GRCh38
5360727 c.1710C>T p.G570G Chr.11: 113739256-113739256 on Build GRCh38
4822724 c.1665C>T p.L555L Chr.11: 113739301-113739301 on Build GRCh38
4017935 c.1656G>A p.L552L Chr.11: 113739310-113739310 on Build GRCh38
4017934 c.1685G>A p.R562H Chr.11: 113739281-113739281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748169308 Chr.11: 113739332-113739332 on Build GRCh38
rs756685419 Chr.11: 113739263-113739263 on Build GRCh38
rs755696652 Chr.11: 113739249-113739249 on Build GRCh38
rs766642448 Chr.11: 113739241-113739241 on Build GRCh38
rs142588649 Chr.11: 113739361-113739361 on Build GRCh38
rs746811678 Chr.11: 113739329-113739329 on Build GRCh38
rs745666929 Chr.11: 113739202-113739202 on Build GRCh38
rs753427313 Chr.11: 113739316-113739316 on Build GRCh38
rs375140483 Chr.11: 113739304-113739304 on Build GRCh38
rs146035001 Chr.11: 113739234-113739234 on Build GRCh38
rs75510745 Chr.11: 113739371-113739371 on Build GRCh38
rs770070787 Chr.11: 113739347-113739347 on Build GRCh38
rs201490208 Chr.11: 113739348-113739348 on Build GRCh38
rs758247426 Chr.11: 113739326-113739326 on Build GRCh38
rs776553004 Chr.11: 113739232-113739232 on Build GRCh38
rs745791786 Chr.11: 113739270-113739270 on Build GRCh38
rs878916556 Chr.11: 113739300-113739300 on Build GRCh38
rs1034178279 Chr.11: 113739283-113739283 on Build GRCh38
rs771796548 Chr.11: 113739207-113739207 on Build GRCh38
rs61762985 Chr.11: 113739281-113739281 on Build GRCh38
rs750896352 Chr.11: 113739238-113739238 on Build GRCh38
rs374255129 Chr.11: 113739354-113739354 on Build GRCh38
rs760801501 Chr.11: 113739218-113739218 on Build GRCh38
rs771967314 Chr.11: 113739278-113739278 on Build GRCh38
rs747306007 Chr.11: 113739282-113739282 on Build GRCh38
rs773919044 Chr.11: 113739299-113739299 on Build GRCh38
rs763428319 Chr.11: 113739239-113739239 on Build GRCh38
rs775837986 Chr.11: 113739217-113739217 on Build GRCh38
rs756817719 Chr.11: 113739318-113739318 on Build GRCh38
rs7938417 Chr.11: 113739248-113739248 on Build GRCh38
rs1046325639 Chr.11: 113739275-113739275 on Build GRCh38
rs779994949 Chr.11: 113739328-113739328 on Build GRCh38
rs984670018 Chr.11: 113739204-113739204 on Build GRCh38
rs768822496 Chr.11: 113739222-113739222 on Build GRCh38
rs763655094 Chr.11: 113739314-113739314 on Build GRCh38
rs146253851 Chr.11: 113739301-113739301 on Build GRCh38
rs777431952 Chr.11: 113739257-113739257 on Build GRCh38
rs759902232 Chr.11: 113739312-113739312 on Build GRCh38
rs558344689 Chr.11: 113739297-113739297 on Build GRCh38
rs148427070 Chr.11: 113739355-113739355 on Build GRCh38
rs748369949 Chr.11: 113739346-113739346 on Build GRCh38
rs778777548 Chr.11: 113739196-113739196 on Build GRCh38
rs768784423 Chr.11: 113739287-113739287 on Build GRCh38
rs774852334 Chr.11: 113739358-113739358 on Build GRCh38
rs754312817 Chr.11: 113739397-113739398 on Build GRCh38
rs368290966 Chr.11: 113739322-113739322 on Build GRCh38
rs778834616 Chr.11: 113739265-113739265 on Build GRCh38
rs766810662 Chr.11: 113739303-113739303 on Build GRCh38
rs776873299 Chr.11: 113739291-113739291 on Build GRCh38
rs776331036 Chr.11: 113739387-113739387 on Build GRCh38
rs370705252 Chr.11: 113739296-113739296 on Build GRCh38

View Full Product Details