Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130905243-130905496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBCD1
Entrez Gene ID
84929
Gene Name
fibrinogen C domain containing 1
Gene Aliases
-
UniGene
Hs.133205
Gene Chromosome Location
Chr.9: 130902438-130939068 on Build GRCh38
Transcripts
NM_001145106.1 NM_032843.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252439 c.987C>G p.Y329* Chr.9: 130905373-130905373 on Build GRCh38
3654930 c.1081G>A p.E361K Chr.9: 130905279-130905279 on Build GRCh38
6550530 c.1030T>A p.Y344N Chr.9: 130905330-130905330 on Build GRCh38
1132572 c.1116C>T p.S372S Chr.9: 130905244-130905244 on Build GRCh38
6664000 c.1025C>T p.T342M Chr.9: 130905335-130905335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779905589 Chr.9: 130905302-130905302 on Build GRCh38
rs935971084 Chr.9: 130905425-130905425 on Build GRCh38
rs760363908 Chr.9: 130905420-130905420 on Build GRCh38
rs754923492 Chr.9: 130905354-130905354 on Build GRCh38
rs1026866063 Chr.9: 130905267-130905267 on Build GRCh38
rs777910575 Chr.9: 130905366-130905366 on Build GRCh38
rs761542131 Chr.9: 130905282-130905282 on Build GRCh38
rs765960401 Chr.9: 130905426-130905426 on Build GRCh38
rs752523144 Chr.9: 130905363-130905363 on Build GRCh38
rs201660431 Chr.9: 130905323-130905323 on Build GRCh38
rs376571211 Chr.9: 130905453-130905453 on Build GRCh38
rs770979473 Chr.9: 130905372-130905372 on Build GRCh38
rs369364369 Chr.9: 130905364-130905364 on Build GRCh38
rs1008252340 Chr.9: 130905392-130905392 on Build GRCh38
rs756525814 Chr.9: 130905342-130905342 on Build GRCh38
rs760182396 Chr.9: 130905318-130905318 on Build GRCh38
rs776937789 Chr.9: 130905448-130905448 on Build GRCh38
rs202150215 Chr.9: 130905319-130905319 on Build GRCh38
rs370497160 Chr.9: 130905369-130905369 on Build GRCh38
rs763941092 Chr.9: 130905449-130905449 on Build GRCh38
rs138622834 Chr.9: 130905307-130905307 on Build GRCh38
rs745883155 Chr.9: 130905380-130905380 on Build GRCh38
rs267602143 Chr.9: 130905403-130905403 on Build GRCh38
rs760142439 Chr.9: 130905338-130905338 on Build GRCh38
rs935671677 Chr.9: 130905292-130905292 on Build GRCh38
rs917115169 Chr.9: 130905265-130905265 on Build GRCh38
rs150732758 Chr.9: 130905334-130905334 on Build GRCh38
rs776098735 Chr.9: 130905386-130905386 on Build GRCh38
rs771501407 Chr.9: 130905432-130905434 on Build GRCh38
rs145939538 Chr.9: 130905306-130905306 on Build GRCh38
rs369214971 Chr.9: 130905462-130905462 on Build GRCh38
rs765909740 Chr.9: 130905346-130905346 on Build GRCh38
rs924429722 Chr.9: 130905404-130905404 on Build GRCh38
rs377608283 Chr.9: 130905309-130905309 on Build GRCh38
rs759010206 Chr.9: 130905427-130905427 on Build GRCh38
rs374267794 Chr.9: 130905273-130905273 on Build GRCh38
rs757511669 Chr.9: 130905461-130905461 on Build GRCh38
rs760023132 Chr.9: 130905269-130905269 on Build GRCh38
rs764246848 Chr.9: 130905409-130905409 on Build GRCh38
rs766829772 Chr.9: 130905262-130905262 on Build GRCh38
rs201321894 Chr.9: 130905428-130905428 on Build GRCh38
rs538234550 Chr.9: 130905325-130905325 on Build GRCh38
rs747883677 Chr.9: 130905310-130905310 on Build GRCh38
rs1052381227 Chr.9: 130905351-130905351 on Build GRCh38
rs73656069 Chr.9: 130905439-130905439 on Build GRCh38
rs373685722 Chr.9: 130905397-130905397 on Build GRCh38
rs753831530 Chr.9: 130905349-130905349 on Build GRCh38
rs563360482 Chr.9: 130905324-130905324 on Build GRCh38
rs367969973 Chr.9: 130905430-130905430 on Build GRCh38
rs770426703 Chr.9: 130905384-130905384 on Build GRCh38
rs376241321 Chr.9: 130905335-130905335 on Build GRCh38
rs781405353 Chr.9: 130905373-130905373 on Build GRCh38
rs373810288 Chr.9: 130905331-130905331 on Build GRCh38
rs772807818 Chr.9: 130905414-130905414 on Build GRCh38
rs903061326 Chr.9: 130905275-130905275 on Build GRCh38
rs767137874 Chr.9: 130905291-130905291 on Build GRCh38
rs139158805 Chr.9: 130905396-130905396 on Build GRCh38
rs778758714 Chr.9: 130905359-130905359 on Build GRCh38
rs747219824 Chr.9: 130905367-130905367 on Build GRCh38
rs1018622669 Chr.9: 130905315-130905315 on Build GRCh38
rs141651387 Chr.9: 130905328-130905328 on Build GRCh38
rs777259026 Chr.9: 130905266-130905266 on Build GRCh38
rs755609942 Chr.9: 130905295-130905295 on Build GRCh38
rs758163576 Chr.9: 130905446-130905446 on Build GRCh38
rs751319931 Chr.9: 130905274-130905274 on Build GRCh38

View Full Product Details