Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 239053387-239053660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC4
Entrez Gene ID
9759
Gene Name
histone deacetylase 4
Gene Aliases
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
UniGene
Hs.20516
Gene Chromosome Location
Chr.2: 239048168-239401647 on Build GRCh38
Transcripts
NM_006037.3 XM_006712877.3 XM_006712878.3 XM_006712879.3 XM_006712880.3 XM_011512217.2 XM_011512218.2 XM_011512219.2 XM_011512220.2 XM_011512221.1 XM_011512222.2 XM_011512223.2 XM_011512224.2 XM_011512225.2 XM_011512226.2 XM_011512227.2 XM_011512230.1 XM_017005394.1 XM_017005395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5715101 c.3196G>A p.V1066M Chr.2: 239053479-239053479 on Build GRCh38
3743708 c.3198G>C p.V1066V Chr.2: 239053477-239053477 on Build GRCh38
5502647 c.3193G>C p.G1065R Chr.2: 239053482-239053482 on Build GRCh38
3972137 c.3145G>A p.E1049K Chr.2: 239053530-239053530 on Build GRCh38
5734782 c.3091_3113del23 p.L1031fs*37 Chr.2: 239053562-239053584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781413370 Chr.2: 239053493-239053493 on Build GRCh38
rs774333411 Chr.2: 239053531-239053531 on Build GRCh38
rs769500122 Chr.2: 239053636-239053636 on Build GRCh38
rs1026813786 Chr.2: 239053418-239053418 on Build GRCh38
rs780193624 Chr.2: 239053577-239053577 on Build GRCh38
rs1056969628 Chr.2: 239053558-239053558 on Build GRCh38
rs775149364 Chr.2: 239053451-239053451 on Build GRCh38
rs772021150 Chr.2: 239053607-239053607 on Build GRCh38
rs371413159 Chr.2: 239053437-239053437 on Build GRCh38
rs753027270 Chr.2: 239053480-239053480 on Build GRCh38
rs764736734 Chr.2: 239053486-239053486 on Build GRCh38
rs145017202 Chr.2: 239053468-239053468 on Build GRCh38
rs375432624 Chr.2: 239053471-239053471 on Build GRCh38
rs750488115 Chr.2: 239053572-239053572 on Build GRCh38
rs765593997 Chr.2: 239053633-239053633 on Build GRCh38
rs771988784 Chr.2: 239053637-239053637 on Build GRCh38
rs781613756 Chr.2: 239053447-239053447 on Build GRCh38
rs891427561 Chr.2: 239053613-239053613 on Build GRCh38
rs747881579 Chr.2: 239053596-239053596 on Build GRCh38
rs778378070 Chr.2: 239053441-239053441 on Build GRCh38
rs115552422 Chr.2: 239053470-239053470 on Build GRCh38
rs766000387 Chr.2: 239053479-239053479 on Build GRCh38
rs769693243 Chr.2: 239053449-239053449 on Build GRCh38
rs756189766 Chr.2: 239053576-239053576 on Build GRCh38
rs765981179 Chr.2: 239053551-239053551 on Build GRCh38
rs754080436 Chr.2: 239053589-239053589 on Build GRCh38
rs746539719 Chr.2: 239053626-239053626 on Build GRCh38
rs768420939 Chr.2: 239053525-239053525 on Build GRCh38
rs200713014 Chr.2: 239053442-239053442 on Build GRCh38
rs114003127 Chr.2: 239053510-239053510 on Build GRCh38
rs760184285 Chr.2: 239053546-239053546 on Build GRCh38
rs763458991 Chr.2: 239053557-239053557 on Build GRCh38
rs371369219 Chr.2: 239053467-239053467 on Build GRCh38
rs374211203 Chr.2: 239053433-239053433 on Build GRCh38
rs992289919 Chr.2: 239053415-239053415 on Build GRCh38
rs200726253 Chr.2: 239053485-239053485 on Build GRCh38
rs745890707 Chr.2: 239053497-239053497 on Build GRCh38
rs776314282 Chr.2: 239053556-239053556 on Build GRCh38
rs778685101 Chr.2: 239053595-239053595 on Build GRCh38
rs771931235 Chr.2: 239053532-239053532 on Build GRCh38
rs757840092 Chr.2: 239053563-239053563 on Build GRCh38
rs774306389 Chr.2: 239053628-239053635 on Build GRCh38
rs754838487 Chr.2: 239053590-239053590 on Build GRCh38
rs772611799 Chr.2: 239053545-239053545 on Build GRCh38
rs759257962 Chr.2: 239053635-239053635 on Build GRCh38
rs768075956 Chr.2: 239053565-239053565 on Build GRCh38
rs764540172 Chr.2: 239053561-239053561 on Build GRCh38
rs752178388 Chr.2: 239053562-239053562 on Build GRCh38
rs770450763 Chr.2: 239053630-239053630 on Build GRCh38
rs777728549 Chr.2: 239053618-239053618 on Build GRCh38
rs752090163 Chr.2: 239053492-239053492 on Build GRCh38
rs767200219 Chr.2: 239053426-239053426 on Build GRCh38
rs142279745 Chr.2: 239053516-239053516 on Build GRCh38
rs756300642 Chr.2: 239053504-239053504 on Build GRCh38
rs201823646 Chr.2: 239053438-239053438 on Build GRCh38
rs750059033 Chr.2: 239053432-239053432 on Build GRCh38
rs749051152 Chr.2: 239053453-239053453 on Build GRCh38

View Full Product Details