Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103300177-103300402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
UniGene
Hs.591863
Gene Chromosome Location
Chr.8: 103298433-103332866 on Build GRCh38
Transcripts
NM_001164615.1 NM_001164616.1 NM_001317796.1 NM_003506.3 NR_133921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668172 c.159T>C p.I53I Chr.8: 103300266-103300266 on Build GRCh38
3884263 c.130A>G p.N44D Chr.8: 103300237-103300237 on Build GRCh38
1453718 c.97A>G p.M33V Chr.8: 103300204-103300204 on Build GRCh38
1453719 c.121delT p.F42fs*4 Chr.8: 103300228-103300228 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs111920622 Chr.8: 103300208-103300208 on Build GRCh38
rs752048729 Chr.8: 103300202-103300202 on Build GRCh38
rs973457330 Chr.8: 103300356-103300357 on Build GRCh38
rs3736046 Chr.8: 103300303-103300303 on Build GRCh38
rs753953103 Chr.8: 103300261-103300261 on Build GRCh38
rs370879185 Chr.8: 103300327-103300327 on Build GRCh38
rs779974159 Chr.8: 103300307-103300307 on Build GRCh38
rs779056600 Chr.8: 103300265-103300265 on Build GRCh38
rs144185033 Chr.8: 103300272-103300272 on Build GRCh38
rs773146626 Chr.8: 103300236-103300236 on Build GRCh38
rs963465921 Chr.8: 103300289-103300289 on Build GRCh38
rs539738840 Chr.8: 103300332-103300332 on Build GRCh38
rs553335677 Chr.8: 103300238-103300238 on Build GRCh38
rs761443232 Chr.8: 103300333-103300333 on Build GRCh38
rs374556380 Chr.8: 103300206-103300206 on Build GRCh38
rs827528 Chr.8: 103300204-103300204 on Build GRCh38
rs571735437 Chr.8: 103300271-103300271 on Build GRCh38
rs746328732 Chr.8: 103300218-103300218 on Build GRCh38
rs768386963 Chr.8: 103300324-103300324 on Build GRCh38
rs773784932 Chr.8: 103300319-103300319 on Build GRCh38
rs768184791 Chr.8: 103300242-103300242 on Build GRCh38
rs970993467 Chr.8: 103300207-103300207 on Build GRCh38
rs777814122 Chr.8: 103300250-103300250 on Build GRCh38
rs746734173 Chr.8: 103300313-103300313 on Build GRCh38
rs757230055 Chr.8: 103300262-103300262 on Build GRCh38
rs752962628 Chr.8: 103300220-103300220 on Build GRCh38

View Full Product Details