Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70530034-70530259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALD1
Entrez Gene ID
27143
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
KIAA1274, PALD
UniGene
Hs.202351
Gene Chromosome Location
Chr.10: 70458542-70568450 on Build GRCh38
Transcripts
NM_014431.2 XM_011539637.2 XM_011539638.2 XM_017016072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
685472 c.442C>T p.Q148* Chr.10: 70530042-70530042 on Build GRCh38
1348837 c.460G>A p.G154R Chr.10: 70530060-70530060 on Build GRCh38
5469433 c.459C>T p.D153D Chr.10: 70530059-70530059 on Build GRCh38
5492662 c.434G>A p.R145Q Chr.10: 70530034-70530034 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897917963 Chr.10: 70530064-70530064 on Build GRCh38
rs751044363 Chr.10: 70530069-70530069 on Build GRCh38
rs1022012161 Chr.10: 70530209-70530209 on Build GRCh38
rs755842979 Chr.10: 70530101-70530101 on Build GRCh38
rs763297834 Chr.10: 70530149-70530149 on Build GRCh38
rs899726028 Chr.10: 70530203-70530203 on Build GRCh38
rs376904569 Chr.10: 70530091-70530091 on Build GRCh38
rs765648109 Chr.10: 70530054-70530054 on Build GRCh38
rs753237838 Chr.10: 70530131-70530131 on Build GRCh38
rs766850114 Chr.10: 70530062-70530062 on Build GRCh38
rs892172181 Chr.10: 70530174-70530174 on Build GRCh38
rs141285632 Chr.10: 70530059-70530059 on Build GRCh38
rs527822719 Chr.10: 70530165-70530165 on Build GRCh38
rs749098092 Chr.10: 70530116-70530116 on Build GRCh38
rs1018226827 Chr.10: 70530196-70530196 on Build GRCh38
rs975193130 Chr.10: 70530225-70530225 on Build GRCh38
rs143550137 Chr.10: 70530060-70530060 on Build GRCh38
rs1007551643 Chr.10: 70530158-70530158 on Build GRCh38
rs963778921 Chr.10: 70530212-70530212 on Build GRCh38
rs754517859 Chr.10: 70530086-70530086 on Build GRCh38
rs948166677 Chr.10: 70530102-70530102 on Build GRCh38
rs777622492 Chr.10: 70530110-70530110 on Build GRCh38
rs552490137 Chr.10: 70530122-70530122 on Build GRCh38
rs34609410 Chr.10: 70530069-70530070 on Build GRCh38
rs906768831 Chr.10: 70530128-70530129 on Build GRCh38
rs767112389 Chr.10: 70530087-70530087 on Build GRCh38
rs1049534400 Chr.10: 70530148-70530148 on Build GRCh38
rs1003828451 Chr.10: 70530135-70530135 on Build GRCh38

View Full Product Details