Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15630200-15630453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDI2
Entrez Gene ID
84301
Gene Name
DNA damage inducible 1 homolog 2
Gene Aliases
-
UniGene
Hs.718857
Gene Chromosome Location
Chr.1: 15617458-15661057 on Build GRCh38
Transcripts
NM_032341.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4908459 c.382T>G p.S128A Chr.1: 15630438-15630438 on Build GRCh38
4891531 c.328C>A p.R110S Chr.1: 15630384-15630384 on Build GRCh38
898044 c.352T>G p.S118A Chr.1: 15630408-15630408 on Build GRCh38
3984373 c.288C>T p.F96F Chr.1: 15630344-15630344 on Build GRCh38
2084672 c.323G>A p.R108Q Chr.1: 15630379-15630379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024852111 Chr.1: 15630244-15630244 on Build GRCh38
rs148633078 Chr.1: 15630385-15630385 on Build GRCh38
rs746374554 Chr.1: 15630406-15630406 on Build GRCh38
rs532223617 Chr.1: 15630412-15630412 on Build GRCh38
rs10927817 Chr.1: 15630252-15630252 on Build GRCh38
rs371977898 Chr.1: 15630372-15630372 on Build GRCh38
rs757198316 Chr.1: 15630409-15630409 on Build GRCh38
rs965858594 Chr.1: 15630226-15630226 on Build GRCh38
rs564313462 Chr.1: 15630378-15630378 on Build GRCh38
rs765246366 Chr.1: 15630269-15630269 on Build GRCh38
rs201289209 Chr.1: 15630299-15630299 on Build GRCh38
rs113104115 Chr.1: 15630374-15630374 on Build GRCh38
rs768561475 Chr.1: 15630279-15630279 on Build GRCh38
rs777823398 Chr.1: 15630278-15630278 on Build GRCh38
rs530424865 Chr.1: 15630247-15630247 on Build GRCh38
rs752896869 Chr.1: 15630295-15630295 on Build GRCh38
rs72647120 Chr.1: 15630274-15630274 on Build GRCh38
rs972334868 Chr.1: 15630250-15630250 on Build GRCh38
rs140092977 Chr.1: 15630379-15630379 on Build GRCh38
rs748799263 Chr.1: 15630338-15630338 on Build GRCh38
rs764221560 Chr.1: 15630400-15630400 on Build GRCh38
rs757729580 Chr.1: 15630384-15630384 on Build GRCh38
rs919161439 Chr.1: 15630272-15630272 on Build GRCh38
rs535427500 Chr.1: 15630343-15630343 on Build GRCh38
rs768110369 Chr.1: 15630351-15630351 on Build GRCh38
rs758544170 Chr.1: 15630308-15630308 on Build GRCh38
rs779619064 Chr.1: 15630334-15630334 on Build GRCh38
rs1022554970 Chr.1: 15630413-15630413 on Build GRCh38
rs745657430 Chr.1: 15630314-15630314 on Build GRCh38
rs778915334 Chr.1: 15630299-15630301 on Build GRCh38
rs750222182 Chr.1: 15630415-15630415 on Build GRCh38
rs751503696 Chr.1: 15630284-15630284 on Build GRCh38
rs1009821391 Chr.1: 15630227-15630227 on Build GRCh38
rs773869764 Chr.1: 15630355-15630355 on Build GRCh38
rs368388796 Chr.1: 15630321-15630321 on Build GRCh38
rs764466841 Chr.1: 15630376-15630376 on Build GRCh38
rs948393633 Chr.1: 15630344-15630344 on Build GRCh38

View Full Product Details