Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33202440-33202713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A7
Entrez Gene ID
7922
Gene Name
solute carrier family 39 member 7
Gene Aliases
D6S115E, D6S2244E, H2-KE4, HKE4, KE4, RING5, ZIP7
UniGene
Hs.631995
Gene Chromosome Location
Chr.6: 33200826-33204437 on Build GRCh38
Transcripts
NM_001077516.1 NM_001288777.1 NM_006979.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770744 c.561G>A p.E187E Chr.6: 33202591-33202591 on Build GRCh38
5130655 c.627G>A p.G209G Chr.6: 33202657-33202657 on Build GRCh38
4153290 c.916G>A p.A306T Chr.6: 33202676-33202676 on Build GRCh38
7157342 c.803G>A p.R268H Chr.6: 33202563-33202563 on Build GRCh38
164407 c.832G>C p.E278Q Chr.6: 33202592-33202592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755621854 Chr.6: 33202529-33202529 on Build GRCh38
rs1043544733 Chr.6: 33202503-33202503 on Build GRCh38
rs996833612 Chr.6: 33202527-33202527 on Build GRCh38
rs774422746 Chr.6: 33202634-33202634 on Build GRCh38
rs780681928 Chr.6: 33202609-33202610 on Build GRCh38
rs200100279 Chr.6: 33202607-33202607 on Build GRCh38
rs765814917 Chr.6: 33202535-33202535 on Build GRCh38
rs749880129 Chr.6: 33202679-33202681 on Build GRCh38
rs762305080 Chr.6: 33202652-33202652 on Build GRCh38
rs753508771 Chr.6: 33202549-33202549 on Build GRCh38
rs943235103 Chr.6: 33202562-33202562 on Build GRCh38
rs113452803 Chr.6: 33202550-33202550 on Build GRCh38
rs780234206 Chr.6: 33202604-33202604 on Build GRCh38
rs758118690 Chr.6: 33202584-33202584 on Build GRCh38
rs199527989 Chr.6: 33202513-33202513 on Build GRCh38
rs748028940 Chr.6: 33202564-33202564 on Build GRCh38
rs751776284 Chr.6: 33202675-33202675 on Build GRCh38
rs758951812 Chr.6: 33202685-33202685 on Build GRCh38
rs202062254 Chr.6: 33202668-33202668 on Build GRCh38
rs746877526 Chr.6: 33202596-33202596 on Build GRCh38
rs1026743380 Chr.6: 33202628-33202628 on Build GRCh38
rs753388445 Chr.6: 33202676-33202676 on Build GRCh38
rs1048778 Chr.6: 33202599-33202599 on Build GRCh38
rs901809244 Chr.6: 33202575-33202575 on Build GRCh38
rs778401742 Chr.6: 33202563-33202563 on Build GRCh38
rs755259564 Chr.6: 33202687-33202687 on Build GRCh38
rs1016376865 Chr.6: 33202689-33202689 on Build GRCh38
rs570973108 Chr.6: 33202629-33202629 on Build GRCh38
rs761117933 Chr.6: 33202669-33202669 on Build GRCh38
rs768127403 Chr.6: 33202479-33202479 on Build GRCh38
rs762061076 Chr.6: 33202466-33202466 on Build GRCh38
rs777925468 Chr.6: 33202585-33202585 on Build GRCh38
rs772352401 Chr.6: 33202655-33202655 on Build GRCh38

View Full Product Details