Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1744896-1745169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINF2
Entrez Gene ID
5345
Gene Name
serpin family F member 2
Gene Aliases
A2AP, AAP, ALPHA-2-PI, API, PLI
UniGene
Hs.159509
Gene Chromosome Location
Chr.17: 1742808-1755265 on Build GRCh38
Transcripts
NM_000934.3 NM_001165920.1 NM_001165921.1 XM_005256701.4 XM_017024765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6435086 c.28C>A p.L10I Chr.17: 1745023-1745023 on Build GRCh38
975970 c.6G>A p.A2A Chr.17: 1745001-1745001 on Build GRCh38
5653362 c.57C>T p.C19C Chr.17: 1745052-1745052 on Build GRCh38
4634160 c.15G>T p.W5C Chr.17: 1745010-1745010 on Build GRCh38
3691382 c.4G>A p.A2T Chr.17: 1744999-1744999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377055860 Chr.17: 1744992-1744992 on Build GRCh38
rs746320572 Chr.17: 1745068-1745068 on Build GRCh38
rs767370481 Chr.17: 1745031-1745031 on Build GRCh38
rs914737056 Chr.17: 1744922-1744922 on Build GRCh38
rs369804932 Chr.17: 1745113-1745113 on Build GRCh38
rs536866023 Chr.17: 1744923-1744923 on Build GRCh38
rs976043138 Chr.17: 1745050-1745050 on Build GRCh38
rs752464909 Chr.17: 1745037-1745037 on Build GRCh38
rs914849811 Chr.17: 1745112-1745112 on Build GRCh38
rs759893506 Chr.17: 1744964-1744964 on Build GRCh38
rs769466657 Chr.17: 1745142-1745142 on Build GRCh38
rs926583692 Chr.17: 1744928-1744928 on Build GRCh38
rs373510687 Chr.17: 1744944-1744944 on Build GRCh38
rs764831217 Chr.17: 1744980-1744980 on Build GRCh38
rs370280422 Chr.17: 1745055-1745055 on Build GRCh38
rs544596507 Chr.17: 1744927-1744927 on Build GRCh38
rs893852610 Chr.17: 1745140-1745140 on Build GRCh38
rs551505236 Chr.17: 1745081-1745081 on Build GRCh38
rs372981803 Chr.17: 1745056-1745056 on Build GRCh38
rs761481781 Chr.17: 1744979-1744979 on Build GRCh38
rs2070862 Chr.17: 1745000-1745000 on Build GRCh38
rs531196259 Chr.17: 1744925-1744925 on Build GRCh38
rs777871727 Chr.17: 1744953-1744953 on Build GRCh38
rs751614553 Chr.17: 1745016-1745016 on Build GRCh38
rs17852960 Chr.17: 1744999-1744999 on Build GRCh38
rs373385252 Chr.17: 1744956-1744956 on Build GRCh38
rs779797130 Chr.17: 1745045-1745045 on Build GRCh38
rs772352419 Chr.17: 1744967-1744967 on Build GRCh38
rs980330383 Chr.17: 1745115-1745115 on Build GRCh38
rs565475587 Chr.17: 1745054-1745054 on Build GRCh38
rs201320264 Chr.17: 1744948-1744948 on Build GRCh38
rs956245824 Chr.17: 1745074-1745074 on Build GRCh38
rs754979455 Chr.17: 1745024-1745024 on Build GRCh38
rs766407419 Chr.17: 1745008-1745008 on Build GRCh38
rs781139539 Chr.17: 1744947-1744947 on Build GRCh38
rs1018100074 Chr.17: 1745027-1745027 on Build GRCh38
rs771450051 Chr.17: 1744960-1744960 on Build GRCh38
rs71359127 Chr.17: 1745106-1745106 on Build GRCh38
rs752826496 Chr.17: 1744924-1744924 on Build GRCh38
rs34712321 Chr.17: 1744939-1744940 on Build GRCh38
rs373572476 Chr.17: 1744943-1744943 on Build GRCh38
rs775794693 Chr.17: 1745097-1745097 on Build GRCh38
rs775841802 Chr.17: 1744972-1744972 on Build GRCh38
rs887589788 Chr.17: 1744988-1744988 on Build GRCh38
rs747375857 Chr.17: 1745139-1745139 on Build GRCh38

View Full Product Details