Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177455895-177456126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR7
Entrez Gene ID
80758
Gene Name
proline rich 7 (synaptic)
Gene Aliases
-
UniGene
Hs.534492
Gene Chromosome Location
Chr.5: 177445936-177456701 on Build GRCh38
Transcripts
NM_001174101.1 NM_001174102.1 NM_030567.4 XM_011534662.1 XM_011534663.2 XM_017009896.1 XM_017009897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5630593 c.630C>T p.Y210Y Chr.5: 177455926-177455926 on Build GRCh38
7316352 c.687C>T p.L229L Chr.5: 177455983-177455983 on Build GRCh38
5732575 c.726C>A p.S242R Chr.5: 177456022-177456022 on Build GRCh38
1292098 c.722C>A p.P241H Chr.5: 177456018-177456018 on Build GRCh38
5787151 c.745A>C p.S249R Chr.5: 177456041-177456041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905045574 Chr.5: 177455932-177455932 on Build GRCh38
rs756956664 Chr.5: 177455920-177455920 on Build GRCh38
rs762162441 Chr.5: 177455985-177455985 on Build GRCh38
rs780982893 Chr.5: 177456025-177456025 on Build GRCh38
rs1034972674 Chr.5: 177455989-177455989 on Build GRCh38
rs770169312 Chr.5: 177456091-177456091 on Build GRCh38
rs958268338 Chr.5: 177455955-177455955 on Build GRCh38
rs781089506 Chr.5: 177455941-177455941 on Build GRCh38
rs777659875 Chr.5: 177456049-177456049 on Build GRCh38
rs200958504 Chr.5: 177456073-177456073 on Build GRCh38
rs960306205 Chr.5: 177456037-177456037 on Build GRCh38
rs755983632 Chr.5: 177456035-177456035 on Build GRCh38
rs1003854231 Chr.5: 177455940-177455940 on Build GRCh38
rs560102319 Chr.5: 177456070-177456070 on Build GRCh38
rs866898403 Chr.5: 177456023-177456023 on Build GRCh38
rs982699207 Chr.5: 177456028-177456028 on Build GRCh38
rs777009396 Chr.5: 177456079-177456079 on Build GRCh38
rs868688427 Chr.5: 177456047-177456047 on Build GRCh38
rs754125537 Chr.5: 177455917-177455917 on Build GRCh38
rs866647625 Chr.5: 177456074-177456074 on Build GRCh38
rs764856740 Chr.5: 177455921-177455921 on Build GRCh38
rs771342319 Chr.5: 177455950-177455950 on Build GRCh38
rs377313231 Chr.5: 177455959-177455959 on Build GRCh38
rs780233186 Chr.5: 177456061-177456061 on Build GRCh38
rs755000520 Chr.5: 177456022-177456022 on Build GRCh38
rs775395167 Chr.5: 177455967-177455967 on Build GRCh38
rs761041020 Chr.5: 177455914-177455914 on Build GRCh38
rs769025501 Chr.5: 177455977-177455977 on Build GRCh38
rs746126934 Chr.5: 177456053-177456053 on Build GRCh38
rs779484358 Chr.5: 177455934-177455934 on Build GRCh38
rs754838997 Chr.5: 177455939-177455939 on Build GRCh38
rs777112365 Chr.5: 177455982-177455982 on Build GRCh38
rs762252364 Chr.5: 177456085-177456085 on Build GRCh38
rs200498552 Chr.5: 177455963-177455963 on Build GRCh38
rs761287941 Chr.5: 177455968-177455968 on Build GRCh38
rs370926335 Chr.5: 177455996-177455996 on Build GRCh38
rs1025603710 Chr.5: 177456077-177456077 on Build GRCh38
rs762551015 Chr.5: 177456004-177456004 on Build GRCh38
rs773532633 Chr.5: 177456097-177456097 on Build GRCh38
rs757909291 Chr.5: 177455927-177455927 on Build GRCh38
rs750092133 Chr.5: 177456001-177456001 on Build GRCh38
rs747050864 Chr.5: 177456066-177456066 on Build GRCh38
rs765953103 Chr.5: 177456010-177456010 on Build GRCh38
rs747882251 Chr.5: 177455943-177455943 on Build GRCh38
rs746997004 Chr.5: 177455938-177455938 on Build GRCh38
rs752610362 Chr.5: 177456033-177456033 on Build GRCh38
rs750003754 Chr.5: 177455922-177455922 on Build GRCh38
rs746026474 Chr.5: 177455954-177455954 on Build GRCh38
rs751010159 Chr.5: 177456020-177456020 on Build GRCh38
rs35777704 Chr.5: 177455969-177455969 on Build GRCh38

View Full Product Details