Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 103537251-103537476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNPC3
Entrez Gene ID
55599
Gene Name
RNA binding region (RNP1, RRM) containing 3
Gene Aliases
RBM40, RNP, SNRNP65
UniGene
Hs.632423
Gene Chromosome Location
Chr.1: 103525956-103555239 on Build GRCh38
Transcripts
NM_017619.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6429971 c.689C>A p.P230H Chr.1: 103537406-103537406 on Build GRCh38
6394296 c.650C>T p.A217V Chr.1: 103537367-103537367 on Build GRCh38
6988222 c.703G>T p.E235* Chr.1: 103537420-103537420 on Build GRCh38
6407012 c.650C>A p.A217E Chr.1: 103537367-103537367 on Build GRCh38
5679289 c.666A>G p.T222T Chr.1: 103537383-103537383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373065876 Chr.1: 103537419-103537419 on Build GRCh38
rs772555074 Chr.1: 103537309-103537309 on Build GRCh38
rs1022046361 Chr.1: 103537435-103537435 on Build GRCh38
rs892303941 Chr.1: 103537347-103537347 on Build GRCh38
rs1030647114 Chr.1: 103537284-103537284 on Build GRCh38
rs759066025 Chr.1: 103537390-103537390 on Build GRCh38
rs749762010 Chr.1: 103537420-103537420 on Build GRCh38
rs547635374 Chr.1: 103537312-103537312 on Build GRCh38
rs573258565 Chr.1: 103537422-103537422 on Build GRCh38
rs376314663 Chr.1: 103537436-103537436 on Build GRCh38
rs760337894 Chr.1: 103537343-103537343 on Build GRCh38
rs370588175 Chr.1: 103537382-103537382 on Build GRCh38
rs748592772 Chr.1: 103537307-103537307 on Build GRCh38
rs566265700 Chr.1: 103537370-103537370 on Build GRCh38
rs539692595 Chr.1: 103537383-103537383 on Build GRCh38
rs868855128 Chr.1: 103537428-103537428 on Build GRCh38
rs554345333 Chr.1: 103537285-103537299 on Build GRCh38

View Full Product Details