Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53859979-53860242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIPF1
Entrez Gene ID
54432
Gene Name
Yip1 domain family member 1
Gene Aliases
DJ167A19.1, FinGER1
UniGene
Hs.11923
Gene Chromosome Location
Chr.1: 53851719-53889831 on Build GRCh38
Transcripts
NM_018982.4 NR_036639.1 NR_036640.1 NR_135075.1 XM_017001499.1 XM_017001500.1 XM_017001501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311160 c.878C>A p.T293K Chr.1: 53860107-53860107 on Build GRCh38
4239490 c.925C>T p.P309S Chr.1: 53860135-53860135 on Build GRCh38
426464 c.847G>T p.A283S Chr.1: 53860138-53860138 on Build GRCh38
4814416 c.922G>T p.A308S Chr.1: 53860138-53860138 on Build GRCh38
4818645 c.940C>T p.L314F Chr.1: 53860120-53860120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117183913 Chr.1: 53860200-53860200 on Build GRCh38
rs779604820 Chr.1: 53860103-53860103 on Build GRCh38
rs748944185 Chr.1: 53860176-53860176 on Build GRCh38
rs765301018 Chr.1: 53860152-53860152 on Build GRCh38
rs766571443 Chr.1: 53860135-53860135 on Build GRCh38
rs764515015 Chr.1: 53860160-53860160 on Build GRCh38
rs372393274 Chr.1: 53860124-53860124 on Build GRCh38
rs11206230 Chr.1: 53860015-53860015 on Build GRCh38
rs982559573 Chr.1: 53860128-53860128 on Build GRCh38
rs531476459 Chr.1: 53860049-53860049 on Build GRCh38
rs553707100 Chr.1: 53860197-53860197 on Build GRCh38
rs758973672 Chr.1: 53860092-53860092 on Build GRCh38
rs369093623 Chr.1: 53860053-53860053 on Build GRCh38
rs781657448 Chr.1: 53860168-53860168 on Build GRCh38
rs778189609 Chr.1: 53860093-53860093 on Build GRCh38
rs760488021 Chr.1: 53860161-53860162 on Build GRCh38
rs763840145 Chr.1: 53860162-53860162 on Build GRCh38
rs748758376 Chr.1: 53860104-53860104 on Build GRCh38
rs747011693 Chr.1: 53860205-53860205 on Build GRCh38
rs541413210 Chr.1: 53860154-53860154 on Build GRCh38
rs759799125 Chr.1: 53860146-53860146 on Build GRCh38
rs760890531 Chr.1: 53860037-53860037 on Build GRCh38
rs758884746 Chr.1: 53860156-53860156 on Build GRCh38
rs150719469 Chr.1: 53860066-53860066 on Build GRCh38
rs754735152 Chr.1: 53860180-53860180 on Build GRCh38
rs761881714 Chr.1: 53860025-53860025 on Build GRCh38
rs200904837 Chr.1: 53860113-53860113 on Build GRCh38
rs747349449 Chr.1: 53860099-53860099 on Build GRCh38
rs755726716 Chr.1: 53860101-53860101 on Build GRCh38
rs753175243 Chr.1: 53860076-53860076 on Build GRCh38
rs1002133213 Chr.1: 53860065-53860065 on Build GRCh38
rs376875020 Chr.1: 53860029-53860029 on Build GRCh38
rs962817545 Chr.1: 53860182-53860182 on Build GRCh38
rs776807936 Chr.1: 53860139-53860139 on Build GRCh38
rs757585297 Chr.1: 53860162-53860162 on Build GRCh38
rs768676158 Chr.1: 53860020-53860020 on Build GRCh38
rs753721247 Chr.1: 53860203-53860204 on Build GRCh38
rs774452709 Chr.1: 53860023-53860023 on Build GRCh38
rs1052760948 Chr.1: 53860012-53860012 on Build GRCh38
rs774071821 Chr.1: 53860111-53860111 on Build GRCh38
rs546854199 Chr.1: 53860216-53860216 on Build GRCh38
rs17109459 Chr.1: 53860201-53860201 on Build GRCh38
rs765354356 Chr.1: 53860072-53860072 on Build GRCh38
rs149606601 Chr.1: 53860046-53860046 on Build GRCh38
rs775105395 Chr.1: 53860157-53860158 on Build GRCh38
rs974521904 Chr.1: 53860190-53860190 on Build GRCh38
rs138273393 Chr.1: 53860027-53860027 on Build GRCh38
rs200922050 Chr.1: 53860153-53860153 on Build GRCh38
rs772989914 Chr.1: 53860122-53860122 on Build GRCh38
rs751913968 Chr.1: 53860161-53860161 on Build GRCh38
rs753871978 Chr.1: 53860064-53860064 on Build GRCh38

View Full Product Details